missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2,2',6,6'-Tetra-tert-butyl-4,4'-dihydroxybiphenyl 98.0+%, TCI America™
Supplier: TCI America T1807200MG
Specifications
2,2′,6,6′-Tetra-tert-butyl-4,4′-dihydroxybiphenyl | |
186°C | |
C28H42O2 | |
200 mg | |
GSOYMOAPJZYXTB-UHFFFAOYSA-N | |
3,3',5,5'-tetra-tert-butyl-[1,1'-biphenyl]-4,4'-diol | |
67185 | |
≥98.0% (GC) |
128-38-1 | |
Yellow | |
MFCD00069443 | |
2,2′C,6,6′C-Tetra-tert-butyl-[1,1′C-biphenyl]-4,4′C-diol | |
CC(C)(C)C1=CC(=CC(=C1O)C(C)(C)C)C1=CC(=C(O)C(=C1)C(C)(C)C)C(C)(C)C | |
410.64 | |
410.64 | |
Crystalline Powder |
Chemical Identifiers
128-38-1 | |
410.64 | |
GSOYMOAPJZYXTB-UHFFFAOYSA-N | |
67185 | |
CC(C)(C)C1=CC(=CC(=C1O)C(C)(C)C)C1=CC(=C(O)C(=C1)C(C)(C)C)C(C)(C)C |
C28H42O2 | |
MFCD00069443 | |
2,2′C,6,6′C-Tetra-tert-butyl-[1,1′C-biphenyl]-4,4′C-diol | |
3,3',5,5'-tetra-tert-butyl-[1,1'-biphenyl]-4,4'-diol |
Safety and Handling
TSCA : No