Learn More
2,2,6-Trimethyl-1,3-dioxin-4-one, cont. up to ca 6% acetone
CAS: 5394-63-8 | C7H10O3 | 142.154 g/mol
$62.93 - $657.66
Chemical Identifiers
| CAS | 5394-63-8 |
|---|---|
| Molecular Formula | C7H10O3 |
| Molecular Weight (g/mol) | 142.154 |
| MDL Number | MFCD00040468 |
| InChI Key | XFRBXZCBOYNMJP-UHFFFAOYSA-N |
| Synonym | 2,2,6-trimethyl-4h-1,3-dioxin-4-one, 2,2,4-trimethyl-6-keto-1,3-dioxin, diketene acetone adduct, 4h-1,3-dioxin-4-one, 2,2,6-trimethyl, 2,2,6-trimethyl-1,3-dioxen-4-one, 2,2,6-trimethyl-1,3-dioxine-4-one, 2,2,6-trimethyl-1,3 dioxin-4-one, pubchem20364, 2,2,6-trimethyl-4h-1,3-dioxin-4-on, acmc-1akcb |
| PubChem CID | 79368 |
| IUPAC Name | 2,2,6-trimethyl-1,3-dioxin-4-one |
| SMILES | CC1=CC(=O)OC(O1)(C)C |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1425414
|
Thermo Scientific Chemicals
A1425414 |
25 g |
Each for $62.93
|
|
|||||
|
AAA1425422
|
Thermo Scientific Chemicals
A1425422 |
100 g |
Each for $174.25
|
|
|||||
|
AAA1425436
|
Thermo Scientific Chemicals
A1425436 |
500 g |
Each for $657.66
|
|
|||||
Description
2,2,6-Trimethyl-1,3-dioxin-4-one is used in the synthesis of dihydropyrimininones as picomolar sodium iodide symporter inhibitors. It is also used in the synthesis of resorcinol amide Hsp90 Inhibitor AT13387. It is used in the synthesis of acetylketene by flash pyrolysis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2,2,6-Trimethyl-1,3-dioxin-4-one is used in the synthesis of dihydropyrimininones as picomolar sodium iodide symporter inhibitors. It is also used in the synthesis of resorcinol amide Hsp90 Inhibitor AT13387. It is used in the synthesis of acetylketene by flash pyrolysis.
Solubility
Soluble in Chloroform, Methanol. Not miscible in water.
Notes
Store at 4°C. Protect from heat. Store away from oxidizing agents.
Chemical Identifiers
| 5394-63-8 | |
| 142.154 | |
| XFRBXZCBOYNMJP-UHFFFAOYSA-N | |
| 79368 | |
| CC1=CC(=O)OC(O1)(C)C |
| C7H10O3 | |
| MFCD00040468 | |
| 2,2,6-trimethyl-4h-1,3-dioxin-4-one, 2,2,4-trimethyl-6-keto-1,3-dioxin, diketene acetone adduct, 4h-1,3-dioxin-4-one, 2,2,6-trimethyl, 2,2,6-trimethyl-1,3-dioxen-4-one, 2,2,6-trimethyl-1,3-dioxine-4-one, 2,2,6-trimethyl-1,3 dioxin-4-one, pubchem20364, 2,2,6-trimethyl-4h-1,3-dioxin-4-on, acmc-1akcb | |
| 2,2,6-trimethyl-1,3-dioxin-4-one |
Specifications
| 5394-63-8 | |
| 1.094 | |
| 14°C (57°F) | |
| 1.461 | |
| 25 g | |
| 2408 | |
| Soluble in Chloroform,Methanol. Not miscible in water. | |
| CC1=CC(=O)OC(O1)(C)C | |
| 142.154 | |
| 142.15 | |
| 2,2,6-Trimethyl-1,3-dioxin-4-one |
| 7°C | |
| 65°C to 67°C (2 mmHg) | |
| C7H10O3 | |
| MFCD00040468 | |
| UN1993 | |
| 2,2,6-trimethyl-4h-1,3-dioxin-4-one, 2,2,4-trimethyl-6-keto-1,3-dioxin, diketene acetone adduct, 4h-1,3-dioxin-4-one, 2,2,6-trimethyl, 2,2,6-trimethyl-1,3-dioxen-4-one, 2,2,6-trimethyl-1,3-dioxine-4-one, 2,2,6-trimethyl-1,3 dioxin-4-one, pubchem20364, 2,2,6-trimethyl-4h-1,3-dioxin-4-on, acmc-1akcb | |
| XFRBXZCBOYNMJP-UHFFFAOYSA-N | |
| 2,2,6-trimethyl-1,3-dioxin-4-one | |
| 79368 | |
| 94% | |
| cont. up to ≈6% acetone |
Safety and Handling
GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P240-P241-P242-P243-P264b-P280-P303+P361+P353-P305+P351+P338-P370+P378q-P501c
H225-H319-H336
DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber : 226-403-3
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only