missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-(2-Aminophenyl)benzimidazole, 98%, Thermo Scientific Chemicals

$83.10 - $270.27

Chemical Identifiers

CAS 5805-39-0
Molecular Formula C13H11N3
Molecular Weight (g/mol) 209.25
MDL Number MFCD00051496
InChI Key YWNXHTNWOQHFRL-UHFFFAOYSA-N
Synonym 2-2-aminophenyl benzimidazole, 2-1h-benzo d imidazol-2-yl aniline, 2-1h-benzimidazol-2-yl aniline, 2-1h-benzoimidazol-2-yl-phenylamine, 2-1h-1,3-benzodiazol-2-yl aniline, benzenamine, 2-1h-benzimidazol-2-yl, 2-o-aminophenyl benzimidazole, 2-1h-benzo d imidazol-2-yl benzenamine, 2-benzimidazol-2-ylphenylamine
PubChem CID 79869
SMILES NC1=CC=CC=C1C1=NC2=CC=CC=C2N1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0703406
View Documents
Thermo Scientific Chemicals
L0703406
5 g N/A
 
AAL0703414
View Documents
Thermo Scientific Chemicals
L0703414
25 g N/A
 
Description

Description

2-(2-Aminophenyl)benzimidazole is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(2-Aminophenyl)benzimidazole is used as pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications

Chemical Identifiers

5805-39-0
209.25
YWNXHTNWOQHFRL-UHFFFAOYSA-N
79869
C13H11N3
MFCD00051496
2-2-aminophenyl benzimidazole, 2-1h-benzo d imidazol-2-yl aniline, 2-1h-benzimidazol-2-yl aniline, 2-1h-benzoimidazol-2-yl-phenylamine, 2-1h-1,3-benzodiazol-2-yl aniline, benzenamine, 2-1h-benzimidazol-2-yl, 2-o-aminophenyl benzimidazole, 2-1h-benzo d imidazol-2-yl benzenamine, 2-benzimidazol-2-ylphenylamine
NC1=CC=CC=C1C1=NC2=CC=CC=C2N1

Specifications

5805-39-0
C13H11N3
5 g
2-2-aminophenyl benzimidazole, 2-1h-benzo d imidazol-2-yl aniline, 2-1h-benzimidazol-2-yl aniline, 2-1h-benzoimidazol-2-yl-phenylamine, 2-1h-1,3-benzodiazol-2-yl aniline, benzenamine, 2-1h-benzimidazol-2-yl, 2-o-aminophenyl benzimidazole, 2-1h-benzo d imidazol-2-yl benzenamine, 2-benzimidazol-2-ylphenylamine
YWNXHTNWOQHFRL-UHFFFAOYSA-N
2-(1H-1,3-benzodiazol-2-yl)aniline
79869
98%
212°C to 216°C
MFCD00051496
173018
Insoluble in water.
NC1=CC=CC=C1C1=NC2=CC=CC=C2N1
209.25
209.24
2-(2-Aminophenyl)benzimidazole
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H311-H332-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P501c

H302-H315-H318-H335

EINECSNumber : 227-361-9

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only