Learn More
2,2'-Biquinoline, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1090703
Description
2,2'-Biquinoline is used as a reagent for spectrophotometric determination of Cu and as an indicator for titration of organolithium reagents, giving yellow-green charge-transfer complexes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2,2′-Biquinoline is used as a reagent for spectrophotometric determination of Cu and as an indicator for titration of organolithium reagents, giving yellow-green charge-transfer complexes.
Solubility
Sparingly soluble in water (0.001g/L).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Light sensitive.
Specifications
2,2'-Biquinoline | |
193°C to 196°C | |
MFCD00006740 | |
187259 | |
Sparingly soluble in water (0.001g/L). | |
C1=CC=C2C(=C1)C=CC(=N2)C3=NC4=CC=CC=C4C=C3 | |
256.308 | |
256.31 |
119-91-5 | |
C18H12N2 | |
1 g | |
2,2'-biquinoline, cuproin, cuproine, 2,2'-diquinolyl, 2,2'-biquinolyl, 2,2'-quinolyl, 2-2-quinolyl quinoline, 2,2-biquinoline, 2,2-diquinolyl, 2,2;-biquinoline | |
WPTCSQBWLUUYDV-UHFFFAOYSA-N | |
2-quinolin-2-ylquinoline | |
8412 | |
≥98% |
Chemical Identifiers
119-91-5 | |
256.308 | |
WPTCSQBWLUUYDV-UHFFFAOYSA-N | |
8412 | |
C1=CC=C2C(=C1)C=CC(=N2)C3=NC4=CC=CC=C4C=C3 |
C18H12N2 | |
MFCD00006740 | |
2,2'-biquinoline, cuproin, cuproine, 2,2'-diquinolyl, 2,2'-biquinolyl, 2,2'-quinolyl, 2-2-quinolyl quinoline, 2,2-biquinoline, 2,2-diquinolyl, 2,2;-biquinoline | |
2-quinolin-2-ylquinoline |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 204-357-5
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only