missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,2'-Biquinoline, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1090714

 View more versions of this product

Catalog No. AAA1090714


Explore available promotions

Description

Description

2,2'-Biquinoline is used as a reagent for spectrophotometric determination of Cu and as an indicator for titration of organolithium reagents, giving yellow-green charge-transfer complexes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,2′-Biquinoline is used as a reagent for spectrophotometric determination of Cu and as an indicator for titration of organolithium reagents, giving yellow-green charge-transfer complexes.

Solubility
Sparingly soluble in water (0.001g/L).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Light sensitive.
Specifications

Specifications

2,2'-Biquinoline
193°C to 196°C
MFCD00006740
187259
Sparingly soluble in water (0.001g/L).
C1=CC=C2C(=C1)C=CC(=N2)C3=NC4=CC=CC=C4C=C3
256.308
256.31
119-91-5
C18H12N2
25 g
2,2'-biquinoline, cuproin, cuproine, 2,2'-diquinolyl, 2,2'-biquinolyl, 2,2'-quinolyl, 2-2-quinolyl quinoline, 2,2-biquinoline, 2,2-diquinolyl, 2,2;-biquinoline
WPTCSQBWLUUYDV-UHFFFAOYSA-N
2-quinolin-2-ylquinoline
8412
≥98%

Chemical Identifiers

119-91-5
256.308
WPTCSQBWLUUYDV-UHFFFAOYSA-N
8412
C1=CC=C2C(=C1)C=CC(=N2)C3=NC4=CC=CC=C4C=C3
C18H12N2
MFCD00006740
2,2'-biquinoline, cuproin, cuproine, 2,2'-diquinolyl, 2,2'-biquinolyl, 2,2'-quinolyl, 2-2-quinolyl quinoline, 2,2-biquinoline, 2,2-diquinolyl, 2,2;-biquinoline
2-quinolin-2-ylquinoline
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 204-357-5

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only