missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,2'-Biquinoline, 98%, Thermo Scientific Chemicals

$66.82 - $265.86

Chemical Identifiers

CAS 119-91-5
Molecular Formula C18H12N2
Molecular Weight (g/mol) 256.3
MDL Number MFCD00006740
InChI Key WPTCSQBWLUUYDV-UHFFFAOYSA-N
Synonym 2,2'-biquinoline, cuproin, cuproine, 2,2'-diquinolyl, 2,2'-biquinolyl, 2,2'-quinolyl, 2-2-quinolyl quinoline, 2,2-biquinoline, 2,2-diquinolyl, 2,2;-biquinoline
PubChem CID 8412
IUPAC Name 2-quinolin-2-ylquinoline
SMILES C1=CC=C2C(=C1)C=CC(=N2)C3=NC4=CC=CC=C4C=C3
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC106300010
SDS View Documents
Thermo Scientific Chemicals
106300010
1 g Glass bottle
Each for $66.82
 
AC106300050
SDS View Documents
Thermo Scientific Chemicals
106300050
5 g Glass bottle
Each for $265.86
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

119-91-5
256.3
WPTCSQBWLUUYDV-UHFFFAOYSA-N
8412
C1=CC=C2C(=C1)C=CC(=N2)C3=NC4=CC=CC=C4C=C3
C18H12N2
MFCD00006740
2,2'-biquinoline, cuproin, cuproine, 2,2'-diquinolyl, 2,2'-biquinolyl, 2,2'-quinolyl, 2-2-quinolyl quinoline, 2,2-biquinoline, 2,2-diquinolyl, 2,2;-biquinoline
2-quinolin-2-ylquinoline

Specifications

119-91-5
100.0
Yellow to Beige
98%
C18H12N2
1 g
2,2'-biquinoline, cuproin, cuproine, 2,2'-diquinolyl, 2,2'-biquinolyl, 2,2'-quinolyl, 2-2-quinolyl quinoline, 2,2-biquinoline, 2,2-diquinolyl, 2,2;-biquinoline
WPTCSQBWLUUYDV-UHFFFAOYSA-N
2-quinolin-2-ylquinoline
8412
98%
2, 2'-Biquinoline, 98%
97.5
193°C to 196°C
Authentic
Glass bottle
MFCD00006740
23, II, 267
Solubility in water: 1mg/l at 25°C. Other solubilities: very soluble in ethanol,,soluble in ether,acetone,benzene,chloroform
C1=CC=C2C(=C1)C=CC(=N2)C3=NC4=CC=CC=C4C=C3
256.3
256.3
Shiny Flakes
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 204-357-5

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only