Learn More
2,2-Bis[4-(4-aminophenoxy)phenyl]propane, 98%, Thermo Scientific Chemicals
$69.10 - $190.95
Chemical Identifiers
CAS | 13080-86-9 |
---|---|
Molecular Formula | C27H26N2O2 |
Molecular Weight (g/mol) | 410.517 |
MDL Number | MFCD00039152 |
InChI Key | KMKWGXGSGPYISJ-UHFFFAOYSA-N |
Synonym | 2,2-bis 4-4-aminophenoxy phenyl propane, 4,4'-propane-2,2-diylbis 4,1-phenylene bis oxy dianiline, 2,2'-bis 4-aminophenoxyphenyl propane, benzenamine, 4,4'-1-methylethylidene bis 4,1-phenyleneoxy bis, 4,4'-isopropylidenebis 4,1-phenyleneoxy dianiline, 4,4'-4,4'-isopropylidenediphenyl-1,1'-diyldioxy dianiline, bis 4-4-aminophenoxy phenyl dimethyl methane, 4,4'-1-methylethylidene bis 4,1-phenyleneoxy bisbenzenamine |
PubChem CID | 83119 |
IUPAC Name | 4-[4-[2-[4-(4-aminophenoxy)phenyl]propan-2-yl]phenoxy]aniline |
SMILES | CC(C)(C1=CC=C(C=C1)OC2=CC=C(C=C2)N)C3=CC=C(C=C3)OC4=CC=C(C=C4)N |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH5505714
|
Thermo Scientific Chemicals
H5505714 |
25 g |
Each for $69.10
|
|
|||||
AAH5505722
|
Thermo Scientific Chemicals
H5505722 |
100 g |
Each for $190.95
|
|
|||||
Description
It is applied in pharmaceutical industry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is applied in pharmaceutical industry.
Solubility
Very slightly soluble in water. Soluble in acetone, Dimethyl sulfoxide(DMSO).
Notes
Store at room temperature with proper ventilation. Keep away from oxidizing agents.
Chemical Identifiers
13080-86-9 | |
410.517 | |
KMKWGXGSGPYISJ-UHFFFAOYSA-N | |
83119 | |
CC(C)(C1=CC=C(C=C1)OC2=CC=C(C=C2)N)C3=CC=C(C=C3)OC4=CC=C(C=C4)N |
C27H26N2O2 | |
MFCD00039152 | |
2,2-bis 4-4-aminophenoxy phenyl propane, 4,4'-propane-2,2-diylbis 4,1-phenylene bis oxy dianiline, 2,2'-bis 4-aminophenoxyphenyl propane, benzenamine, 4,4'-1-methylethylidene bis 4,1-phenyleneoxy bis, 4,4'-isopropylidenebis 4,1-phenyleneoxy dianiline, 4,4'-4,4'-isopropylidenediphenyl-1,1'-diyldioxy dianiline, bis 4-4-aminophenoxy phenyl dimethyl methane, 4,4'-1-methylethylidene bis 4,1-phenyleneoxy bisbenzenamine | |
4-[4-[2-[4-(4-aminophenoxy)phenyl]propan-2-yl]phenoxy]aniline |
Specifications
13080-86-9 | |
C27H26N2O2 | |
25 g | |
Very slightly soluble in water. Soluble in acetone,Dimethyl sulfoxide(DMSO). | |
CC(C)(C1=CC=C(C=C1)OC2=CC=C(C=C2)N)C3=CC=C(C=C3)OC4=CC=C(C=C4)N | |
410.517 | |
410.52 | |
2,2-Bis[4-(4-aminophenoxy)phenyl]propane |
127°C to 130°C | |
MFCD00039152 | |
2,2-bis 4-4-aminophenoxy phenyl propane, 4,4'-propane-2,2-diylbis 4,1-phenylene bis oxy dianiline, 2,2'-bis 4-aminophenoxyphenyl propane, benzenamine, 4,4'-1-methylethylidene bis 4,1-phenyleneoxy bis, 4,4'-isopropylidenebis 4,1-phenyleneoxy dianiline, 4,4'-4,4'-isopropylidenediphenyl-1,1'-diyldioxy dianiline, bis 4-4-aminophenoxy phenyl dimethyl methane, 4,4'-1-methylethylidene bis 4,1-phenyleneoxy bisbenzenamine | |
KMKWGXGSGPYISJ-UHFFFAOYSA-N | |
4-[4-[2-[4-(4-aminophenoxy)phenyl]propan-2-yl]phenoxy]aniline | |
83119 | |
98% |
Safety and Handling
GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P272-P280g-P301+P312-P302+P352-P330-P333+P313-P363-P501c
H302-H317
EINECSNumber : 235-985-8
RTECSNumber : CY1062500
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only