missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2,2-Bis(4-methylphenyl)hexafluoropropane 98.0+%, TCI America™
Supplier: TCI America B141710G
Specifications
2,2-Bis(4-methylphenyl)hexafluoropropane | |
83°C | |
112°C | |
MFCD00042597 | |
4,4′C-(Hexafluoroisopropylidene)ditoluene | |
CC1=CC=C(C=C1)C(C2=CC=C(C=C2)C)(C(F)(F)F)C(F)(F)F | |
332.289 | |
332.29 | |
Crystalline Powder |
1095-77-8 | |
White-Yellow | |
C17H14F6 | |
10 g | |
OWEIAGSMFHSSES-UHFFFAOYSA-N | |
1-[1,1,1,3,3,3-hexafluoro-2-(4-methylphenyl)propan-2-yl]-4-methylbenzene | |
621931 | |
≥98.0% (GC) |
Chemical Identifiers
1095-77-8 | |
332.289 | |
OWEIAGSMFHSSES-UHFFFAOYSA-N | |
621931 | |
CC1=CC=C(C=C1)C(C2=CC=C(C=C2)C)(C(F)(F)F)C(F)(F)F |
C17H14F6 | |
MFCD00042597 | |
4,4′C-(Hexafluoroisopropylidene)ditoluene | |
1-[1,1,1,3,3,3-hexafluoro-2-(4-methylphenyl)propan-2-yl]-4-methylbenzene |
Safety and Handling
TSCA : No