Learn More
2,2-Di-n-propylacetamide, 97%, Thermo Scientific Chemicals
$178.59 - $580.73
Chemical Identifiers
CAS | 2430-27-5 |
---|---|
Molecular Formula | C8H17NO |
Molecular Weight (g/mol) | 143.23 |
MDL Number | MFCD00051534 |
InChI Key | OMOMUFTZPTXCHP-UHFFFAOYSA-N |
Synonym | valpromide, depamide, dipropylacetamide, valpramide, 2-propylpentamide, 2-propylvaleramide, pentanamide, 2-propyl, 2,2-di-n-propylacetamide, 2-ethylvaleramide, propyl-2 valeramide |
PubChem CID | 71113 |
ChEBI | CHEBI:74562 |
IUPAC Name | 2-propylpentanamide |
SMILES | CCCC(CCC)C(N)=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
AAL0884706
|
Thermo Scientific Chemicals
L0884706 |
5 g |
Each for $178.59
|
|
||||||
AAL0884714
|
Thermo Scientific Chemicals
L0884714 |
25 g |
Each for $580.73
|
|
||||||
Description
It is employed as an antiepileptic drug. It is hydrolyzed quickly to VPA in vivo, but has intrinsic anticonvulsant activity. Also a reactant used in the synthesis of mutual prodrugs containing bio-cleavable and drug releasable disulfide linkers.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
SolubilitySoluble in chloroform, dimethyl sulfoxide and methanol. Insoluble in water.
Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Chemical Identifiers
2430-27-5 | |
143.23 | |
OMOMUFTZPTXCHP-UHFFFAOYSA-N | |
71113 | |
2-propylpentanamide |
C8H17NO | |
MFCD00051534 | |
valpromide, depamide, dipropylacetamide, valpramide, 2-propylpentamide, 2-propylvaleramide, pentanamide, 2-propyl, 2,2-di-n-propylacetamide, 2-ethylvaleramide, propyl-2 valeramide | |
CHEBI:74562 | |
CCCC(CCC)C(N)=O |
Specifications
2430-27-5 | |
Odorless | |
MFCD00051534 | |
1750444 | |
valpromide, depamide, dipropylacetamide, valpramide, 2-propylpentamide, 2-propylvaleramide, pentanamide, 2-propyl, 2,2-di-n-propylacetamide, 2-ethylvaleramide, propyl-2 valeramide | |
OMOMUFTZPTXCHP-UHFFFAOYSA-N | |
2-propylpentanamide | |
71113 | |
143.23 | |
2,2-Di-n-propylacetamide |
123°C to 127°C | |
C8H17NO | |
5 g | |
14,9914 | |
Soluble in chloroform,dimethyl sulfoxide and methanol. Insoluble in water. | |
CCCC(CCC)C(N)=O | |
143.23 | |
CHEBI:74562 | |
97% |
Safety and Handling
GHS H Statement
H302
Harmful if swallowed.
P264b-P270-P301+P312-P330-P501c
H302
EINECSNumber : 219-394-2
RTECSNumber : YV5965500
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only