missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,2'-Dichloro-4,4'-bipyridine, 95%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H27695MC

 View more versions of this product

Catalog No. AAH27695MC


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2,2'-Dichloro-4,4'-bipyridine
240°C to 243°C
MFCD01106345
2,2'-dichloro-4,4'-bipyridine, 2-chloro-4-2-chloropyridin-4-yl pyridine, 2,2'-dichloro-4,4'bipyridine, 2,2-dichloro-4,4-bipyridine, pubchem19451, bpcl37, acmc-1ap45, 4,4'-bi 2-chloropyridine, 2,2'-dichloro-4,4' bipyridine
C1=CN=C(C=C1C2=CC(=NC=C2)Cl)Cl
225.072
225.08
53344-74-4
C10H6Cl2N2
100 mg
KSELKNWXBOXHEV-UHFFFAOYSA-N
2-chloro-4-(2-chloropyridin-4-yl)pyridine
5172097
95%

Chemical Identifiers

53344-74-4
225.072
KSELKNWXBOXHEV-UHFFFAOYSA-N
5172097
C1=CN=C(C=C1C2=CC(=NC=C2)Cl)Cl
C10H6Cl2N2
MFCD01106345
2,2'-dichloro-4,4'-bipyridine, 2-chloro-4-2-chloropyridin-4-yl pyridine, 2,2'-dichloro-4,4'bipyridine, 2,2-dichloro-4,4-bipyridine, pubchem19451, bpcl37, acmc-1ap45, 4,4'-bi 2-chloropyridine, 2,2'-dichloro-4,4' bipyridine
2-chloro-4-(2-chloropyridin-4-yl)pyridine
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only