missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2,2-Diethoxypropane 95.0+%, TCI America™
$134.95 - $996.54
Chemical Identifiers
CAS | 126-84-1 |
---|---|
Molecular Formula | C7H16O2 |
Molecular Weight (g/mol) | 132.203 |
MDL Number | MFCD00009224 |
InChI Key | FGQLGYBGTRHODR-UHFFFAOYSA-N |
Synonym | acetone diethyl acetal, propane, 2,2-diethoxy, acetone diethyl ketal, acetone, diethyl acetal, usaf do-44, unii-18i1qhh2k2, acetonediethylketal, acetone diethylacetal, diethoxypropane, propane,2-diethoxy |
PubChem CID | 31361 |
IUPAC Name | 2,2-diethoxypropane |
SMILES | CCOC(C)(C)OCC |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
D045910ML
|
TCI America
D045910ML |
10 mL |
Each for $134.95
|
|
|||||
D0459250ML
|
TCI America
D0459250ML |
250 mL |
Each for $996.54
|
|
|||||
Chemical Identifiers
126-84-1 | |
132.203 | |
FGQLGYBGTRHODR-UHFFFAOYSA-N | |
31361 | |
CCOC(C)(C)OCC |
C7H16O2 | |
MFCD00009224 | |
acetone diethyl acetal, propane, 2,2-diethoxy, acetone diethyl ketal, acetone, diethyl acetal, usaf do-44, unii-18i1qhh2k2, acetonediethylketal, acetone diethylacetal, diethoxypropane, propane,2-diethoxy | |
2,2-diethoxypropane |
Specifications
126-84-1 | |
113°C | |
MFCD00009224 | |
1993 | |
FGQLGYBGTRHODR-UHFFFAOYSA-N | |
2,2-diethoxypropane | |
31361 | |
≥95.0% (GC) | |
2,2-Diethoxypropane |
Yellow | |
C7H16O2 | |
10 mL | |
acetone diethyl acetal, propane, 2,2-diethoxy, acetone diethyl ketal, acetone, diethyl acetal, usaf do-44, unii-18i1qhh2k2, acetonediethylketal, acetone diethylacetal, diethoxypropane, propane,2-diethoxy | |
CCOC(C)(C)OCC | |
132.203 | |
132.20 | |
Liquid |
Safety and Handling
RTECSNumber : AL4900000
TSCA : Yes
Recommended Storage : Refrigerator