missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,2-Difluoro-4-pentenoic acid, 97%, Thermo Scientific Chemicals

$117.25 - $390.81

Chemical Identifiers

CAS 55039-89-9
Molecular Formula C5H6F2O2
Molecular Weight (g/mol) 136.098
MDL Number MFCD09800642
InChI Key LHOKYUDUAYXFGF-UHFFFAOYSA-N
Synonym 2,2-difluoropent-4-enoicacid, 2,2-difluoro-4-pentenoic acid, 4-pentenoic acid, 2,2-difluoro, 2,2-difluoro-pent-4-enoic acid, 10.14272/lhokyuduayxfgf-uhfffaoysa-n, doi:10.14272/lhokyuduayxfgf-uhfffaoysa-n
PubChem CID 11954656
IUPAC Name 2,2-difluoropent-4-enoic acid
SMILES C=CCC(C(=O)O)(F)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH3217303
View Documents
Thermo Scientific Chemicals
H3217303
1 g
Each for $117.25
 
AAH3217306
View Documents
Thermo Scientific Chemicals
H3217306
5 g
Each for $390.81
 
Description

Description

2,2-Difluoro-4-pentenoic acid is involved in the iodolactonization reaction to give the corresponding gamma- lactone, which is converted to prepare prepare 5-hydroxy-3,3-difluoropiperidine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,2-Difluoro-4-pentenoic acid is involved in the iodolactonization reaction to give the corresponding gamma- lactone, which is converted to prepare prepare 5-hydroxy-3,3-difluoropiperidine.

Solubility
Immiscible with water.

Notes
Incompatible with oxidizing agents and bases.
Specifications

Chemical Identifiers

55039-89-9
136.098
LHOKYUDUAYXFGF-UHFFFAOYSA-N
11954656
C=CCC(C(=O)O)(F)F
C5H6F2O2
MFCD09800642
2,2-difluoropent-4-enoicacid, 2,2-difluoro-4-pentenoic acid, 4-pentenoic acid, 2,2-difluoro, 2,2-difluoro-pent-4-enoic acid, 10.14272/lhokyuduayxfgf-uhfffaoysa-n, doi:10.14272/lhokyuduayxfgf-uhfffaoysa-n
2,2-difluoropent-4-enoic acid

Specifications

55039-89-9
C5H6F2O2
1 g
2,2-difluoropent-4-enoicacid, 2,2-difluoro-4-pentenoic acid, 4-pentenoic acid, 2,2-difluoro, 2,2-difluoro-pent-4-enoic acid, 10.14272/lhokyuduayxfgf-uhfffaoysa-n, doi:10.14272/lhokyuduayxfgf-uhfffaoysa-n
LHOKYUDUAYXFGF-UHFFFAOYSA-N
2,2-difluoropent-4-enoic acid
11954656
97%
56°C to 58°C (3 mmHg)
MFCD09800642
UN3265
Immiscible with water.
C=CCC(C(=O)O)(F)F
136.098
136.1
2,2-Difluoro-4-pentenoic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c

H302-H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only