missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,2-Dimethoxy-2-phenylacetophenone, 99%, Thermo Scientific Chemicals

$129.60 - $129.60

Chemical Identifiers

CAS 24650-42-8
Molecular Formula C16H16O3
Molecular Weight (g/mol) 256.30
MDL Number MFCD00008475
InChI Key KWVGIHKZDCUPEU-UHFFFAOYSA-N
Synonym 2,2-dimethoxy-2-phenylacetophenone, benzil dimethyl ketal, kayacure bdmk, irgacure 651, ethanone, 2,2-dimethoxy-1,2-diphenyl, lucirin bdk, photomer 51, benzil dimethylketal, esacure kb 1, irgacure 621
PubChem CID 90571
IUPAC Name 2,2-dimethoxy-1,2-diphenylethanone
SMILES COC(OC)(C(=O)C1=CC=CC=C1)C1=CC=CC=C1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC187840500
SDS View Documents
Thermo Scientific Chemicals
187840500
50 g Glass bottle
Each for $129.60
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

24650-42-8
256.30
KWVGIHKZDCUPEU-UHFFFAOYSA-N
90571
COC(OC)(C(=O)C1=CC=CC=C1)C1=CC=CC=C1
C16H16O3
MFCD00008475
2,2-dimethoxy-2-phenylacetophenone, benzil dimethyl ketal, kayacure bdmk, irgacure 651, ethanone, 2,2-dimethoxy-1,2-diphenyl, lucirin bdk, photomer 51, benzil dimethylketal, esacure kb 1, irgacure 621
2,2-dimethoxy-1,2-diphenylethanone

Specifications

24650-42-8
100.0
White to Yellow
Authentic
Glass bottle
C6H5COC(OCH3)2C6H5
50 g
2,2-dimethoxy-2-phenylacetophenone, benzil dimethyl ketal, kayacure bdmk, irgacure 651, ethanone, 2,2-dimethoxy-1,2-diphenyl, lucirin bdk, photomer 51, benzil dimethylketal, esacure kb 1, irgacure 621
COC(OC)(C(=O)C1=CC=CC=C1)C1=CC=CC=C1
256.30
256.29
Crystalline Powder
98.5
64°C to 68°C
0.2% max. (75°C, 2 hrs)
98.5% min. (GC)
C16H16O3
MFCD00008475
14, 3392
KWVGIHKZDCUPEU-UHFFFAOYSA-N
2,2-dimethoxy-1,2-diphenylethanone
90571
99%
2, 2-Dimethoxy-2-phenylacetophenone, 99%
Videos
Safety and Handling

Safety and Handling

LIGHT SENSITIVE

EINECSNumber : 246-386-6

RTECSNumber : KM5775658

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only