missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,2-Dimethyl-1,3-dichloropropane 95.0+%, TCI America™
$100.67 - $300.11
Chemical Identifiers
| CAS | 29559-55-5 |
|---|---|
| Molecular Formula | C5H10Cl2 |
| Molecular Weight (g/mol) | 141.035 |
| MDL Number | MFCD00039363 |
| InChI Key | KTWNITKLQPCZSL-UHFFFAOYSA-N |
| Synonym | 1,3-Dichloro-2,2-dimethylpropane, 1,3-Dichloroneopentane |
| PubChem CID | 122408 |
| IUPAC Name | 1,3-dichloro-2,2-dimethylpropane |
| SMILES | CC(C)(CCl)CCl |
Chemical Identifiers
| 29559-55-5 | |
| 141.035 | |
| KTWNITKLQPCZSL-UHFFFAOYSA-N | |
| 122408 | |
| CC(C)(CCl)CCl |
| C5H10Cl2 | |
| MFCD00039363 | |
| 1,3-Dichloro-2,2-dimethylpropane, 1,3-Dichloroneopentane | |
| 1,3-dichloro-2,2-dimethylpropane |
Specifications
| 29559-55-5 | |
| 138°C | |
| MFCD00039363 | |
| 1993 | |
| KTWNITKLQPCZSL-UHFFFAOYSA-N | |
| 1,3-dichloro-2,2-dimethylpropane | |
| 122408 | |
| ≥95.0% (GC) | |
| 2,2-Dimethyl-1,3-dichloropropane |
| Colorless | |
| C5H10Cl2 | |
| 5 g | |
| 1,3-Dichloro-2,2-dimethylpropane, 1,3-Dichloroneopentane | |
| CC(C)(CCl)CCl | |
| 141.035 | |
| 141.04 | |
| Liquid |
Safety and Handling
TSCA : No