missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,2-Dimethyl-2,3-dihydro-1-benzofuran-7-carbaldehyde, 97%, Thermo Scientific™
$85.17 - $239.70
Chemical Identifiers
| CAS | 38002-88-9 |
|---|---|
| Molecular Formula | C11H12O2 |
| Molecular Weight (g/mol) | 176.22 |
| MDL Number | MFCD04115387 |
| InChI Key | HGKYVFOYQUSRQN-UHFFFAOYSA-N |
| Synonym | 2,2-dimethyl-2,3-dihydro-1-benzofuran-7-carbaldehyde, 2,2-dimethyl-2,3-dihydrobenzofuran-7-carbaldehyde, 7-benzofurancarboxaldehyde, 2,3-dihydro-2,2-dimethyl, 2,2-dimethyl-7-formyl-2,3-dihydrobenzofuran, 2,2-dimethyl-2,3-dihydro-benzofuran-7-carbaldehyde, 2,3-dihydro-2,2-dimethylbenzofuran-7-carbaldehyde, 2,2-dimethyl-2,3-dihydrobenzo b furan-7-carbaldehyde, 2,3-dihydro-2,2-dimethylbenzofuran-7-carboxaldehyde, 2,2-dimethyl-2,3-dihydro-1-benzo b furan-7-carbaldehyde |
| PubChem CID | 2795471 |
| SMILES | CC1(C)CC2=CC=CC(C=O)=C2O1 |
Chemical Identifiers
| 38002-88-9 | |
| 176.22 | |
| HGKYVFOYQUSRQN-UHFFFAOYSA-N | |
| 2795471 |
| C11H12O2 | |
| MFCD04115387 | |
| 2,2-dimethyl-2,3-dihydro-1-benzofuran-7-carbaldehyde, 2,2-dimethyl-2,3-dihydrobenzofuran-7-carbaldehyde, 7-benzofurancarboxaldehyde, 2,3-dihydro-2,2-dimethyl, 2,2-dimethyl-7-formyl-2,3-dihydrobenzofuran, 2,2-dimethyl-2,3-dihydro-benzofuran-7-carbaldehyde, 2,3-dihydro-2,2-dimethylbenzofuran-7-carbaldehyde, 2,2-dimethyl-2,3-dihydrobenzo b furan-7-carbaldehyde, 2,3-dihydro-2,2-dimethylbenzofuran-7-carboxaldehyde, 2,2-dimethyl-2,3-dihydro-1-benzo b furan-7-carbaldehyde | |
| CC1(C)CC2=CC=CC(C=O)=C2O1 |
Specifications
| 38002-88-9 | |
| 100.0 | |
| Amber Glass Bottle | |
| MFCD04115387 | |
| 2,2-dimethyl-2,3-dihydro-1-benzofuran-7-carbaldehyde, 2,2-dimethyl-2,3-dihydrobenzofuran-7-carbaldehyde, 7-benzofurancarboxaldehyde, 2,3-dihydro-2,2-dimethyl, 2,2-dimethyl-7-formyl-2,3-dihydrobenzofuran, 2,2-dimethyl-2,3-dihydro-benzofuran-7-carbaldehyde, 2,3-dihydro-2,2-dimethylbenzofuran-7-carbaldehyde, 2,2-dimethyl-2,3-dihydrobenzo b furan-7-carbaldehyde, 2,3-dihydro-2,2-dimethylbenzofuran-7-carboxaldehyde, 2,2-dimethyl-2,3-dihydro-1-benzo b furan-7-carbaldehyde | |
| CC1(C)CC2=CC=CC(C=O)=C2O1 | |
| 176.22 | |
| 176.21 | |
| 2, 2-Dimethyl-2, 3-dihydro-1-benzofuran-7-carbaldehyde |
| 96.5 | |
| 97% | |
| C11H12O2 | |
| 250 mg | |
| HGKYVFOYQUSRQN-UHFFFAOYSA-N | |
| 2,2-dimethyl-2,3-dihydro-1-benzofuran-7-carbaldehyde | |
| 2795471 | |
| 97% |