Learn More
2,2-Dimethyl-4-pentenoic acid, 95%, Thermo Scientific Chemicals
$118.90 - $421.32
Chemical Identifiers
CAS | 16386-93-9 |
---|---|
Molecular Formula | C7H12O2 |
Molecular Weight (g/mol) | 128.17 |
MDL Number | MFCD00012304 |
InChI Key | BGUAPYRHJPWVEM-UHFFFAOYSA-N |
Synonym | 2,2-dimethyl-4-pentenoic acid, 4-pentenoic acid, 2,2-dimethyl, 2,2-dimethyl-pent-4-enoic acid, 2,2-dimethyl-4-pentenoicacid, pubchem12797, acmc-1c5lb, ksc495e3r, 2,2-dimethyl4-pentenoic acid, 2,2-dimethyl-1-4-pentenoic acid 1g |
PubChem CID | 140065 |
IUPAC Name | 2,2-dimethylpent-4-enoic acid |
SMILES | CC(C)(CC=C)C(O)=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH6094003
|
Thermo Scientific Chemicals
H6094003 |
1 g |
Each for $118.90
|
|
|||||
AAH6094006
|
Thermo Scientific Chemicals
H6094006 |
5 g |
Each for $421.32
|
|
|||||
Description
It is used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is used as a pharmaceutical intermediate.
Solubility
Soluble in water.
Notes
Incompatible with strong oxidizing agents. Store under room temperature.
Chemical Identifiers
16386-93-9 | |
128.17 | |
BGUAPYRHJPWVEM-UHFFFAOYSA-N | |
140065 | |
CC(C)(CC=C)C(O)=O |
C7H12O2 | |
MFCD00012304 | |
2,2-dimethyl-4-pentenoic acid, 4-pentenoic acid, 2,2-dimethyl, 2,2-dimethyl-pent-4-enoic acid, 2,2-dimethyl-4-pentenoicacid, pubchem12797, acmc-1c5lb, ksc495e3r, 2,2-dimethyl4-pentenoic acid, 2,2-dimethyl-1-4-pentenoic acid 1g | |
2,2-dimethylpent-4-enoic acid |
Specifications
16386-93-9 | |
104°C to 108°C | |
C7H12O2 | |
H2C=CHCH2C(CH3)2CO2H | |
1 g | |
2,2-dimethyl-4-pentenoic acid, 4-pentenoic acid, 2,2-dimethyl, 2,2-dimethyl-pent-4-enoic acid, 2,2-dimethyl-4-pentenoicacid, pubchem12797, acmc-1c5lb, ksc495e3r, 2,2-dimethyl4-pentenoic acid, 2,2-dimethyl-1-4-pentenoic acid 1g | |
BGUAPYRHJPWVEM-UHFFFAOYSA-N | |
2,2-dimethylpent-4-enoic acid | |
140065 | |
95% |
0.933 | |
88°C (190°F) | |
1.431 | |
MFCD00012304 | |
UN3265 | |
Soluble in water. | |
CC(C)(CC=C)C(O)=O | |
128.17 | |
128.17 | |
2,2-Dimethyl-4-pentenoic acid |
Safety and Handling
GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
P210-P235-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
H227-H302-H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only