missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,2-Dimethyl-6-nitro-2H-1,4-benzoxazin-3(4H)-one, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H61097MD

 View more versions of this product

Catalog No. AAH61097MD


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2,2-Dimethyl-6-nitro-2H-1,4-benzoxazin-3(4H)-one
C10H10N2O4
250 mg
YKXZRZGZJZYBBH-UHFFFAOYSA-N
2,2-dimethyl-6-nitro-4H-1,4-benzoxazin-3-one
10656631
97%
85160-84-5
MFCD03427729
2,2-dimethyl-6-nitro-2h-benzo b 1,4 oxazin-3 4h-one, 2h-1,4-benzoxazin-3 4h-one, 2,2-dimethyl-6-nitro, 2,2-dimethyl-6-nitro-2h-1,4-benzoxazin-3 4h-one, 2,2-dimethyl-6-nitro-4h-benzo 1,4 oxazin-3-one, 2,2-dimethyl-6-nitro-2,3-dihydro-4h-1,4-benzooxazine-3-one, 3,4-dihydro-2,2-dimethyl-6-nitro-3-oxo-2h-1,4-benzoxazine, 2,2-dimethyl-6-nitro-2h-benzo b 1.4 oxazine-3 4h-one, 2,2-dimethyl-6-nitro-3,4-dihydro-2h-benzo 1,4 oxazine-one
CC1(C)OC2=C(NC1=O)C=C(C=C2)[N+]([O-])=O
222.20
222.2

Chemical Identifiers

85160-84-5
222.20
YKXZRZGZJZYBBH-UHFFFAOYSA-N
10656631
CC1(C)OC2=C(NC1=O)C=C(C=C2)[N+]([O-])=O
C10H10N2O4
MFCD03427729
2,2-dimethyl-6-nitro-2h-benzo b 1,4 oxazin-3 4h-one, 2h-1,4-benzoxazin-3 4h-one, 2,2-dimethyl-6-nitro, 2,2-dimethyl-6-nitro-2h-1,4-benzoxazin-3 4h-one, 2,2-dimethyl-6-nitro-4h-benzo 1,4 oxazin-3-one, 2,2-dimethyl-6-nitro-2,3-dihydro-4h-1,4-benzooxazine-3-one, 3,4-dihydro-2,2-dimethyl-6-nitro-3-oxo-2h-1,4-benzoxazine, 2,2-dimethyl-6-nitro-2h-benzo b 1.4 oxazine-3 4h-one, 2,2-dimethyl-6-nitro-3,4-dihydro-2h-benzo 1,4 oxazine-one
2,2-dimethyl-6-nitro-4H-1,4-benzoxazin-3-one
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only