missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,2-Dimethylcyclohexanone, Thermo Scientific™

Chemical Identifiers

CAS 1193-47-1
Molecular Formula C8H14O
Molecular Weight (g/mol) 126.2
InChI Key KNSPBSQWRKKAPI-UHFFFAOYSA-N
Synonym 2,2-dimethylcyclohexanone, cyclohexanone, 2,2-dimethyl, dimethylcyclohexanone, cyclohexanone, dimethyl, 6,6-dimethylcyclohexanone, cyclohexanone,2-dimethyl, acmc-20a0u6, 2,2-dimethyl-cyclohexanone, cyclohexanone,2,2-dimethyl, 2,2-dimethyl-1-cyclohexanone
PubChem CID 136929
IUPAC Name 2,2-dimethylcyclohexan-1-one
SMILES CC1(CCCCC1=O)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Qty  
Catalog Number Mfr. No. Quantity Packaging Price Qty  
AC426500010
SDS missing translation for 'viewDocuments'
Thermo Scientific Chemicals
426500010
1g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC426500050
SDS missing translation for 'viewDocuments'
Thermo Scientific Chemicals
426500050
5g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

1193-47-1
126.2
2,2-dimethylcyclohexanone, cyclohexanone, 2,2-dimethyl, dimethylcyclohexanone, cyclohexanone, dimethyl, 6,6-dimethylcyclohexanone, cyclohexanone,2-dimethyl, acmc-20a0u6, 2,2-dimethyl-cyclohexanone, cyclohexanone,2,2-dimethyl, 2,2-dimethyl-1-cyclohexanone
2,2-dimethylcyclohexan-1-one
C8H14O
KNSPBSQWRKKAPI-UHFFFAOYSA-N
136929
CC1(CCCCC1=O)C

Specifications

1193-47-1
169.0°C to 170.0°C (0.8mmHg)
Authentic
C8H14O
1.4470 to 1.4490 (20°C, 589nm)
0.912
KNSPBSQWRKKAPI-UHFFFAOYSA-N
2,2-dimethylcyclohexan-1-one
136929
≥94%
0.9120g/mL
50°C
Glass bottle
(CH3)2C6H8(=O)
1g
2,2-dimethylcyclohexanone, cyclohexanone, 2,2-dimethyl, dimethylcyclohexanone, cyclohexanone, dimethyl, 6,6-dimethylcyclohexanone, cyclohexanone,2-dimethyl, acmc-20a0u6, 2,2-dimethyl-cyclohexanone, cyclohexanone,2,2-dimethyl, 2,2-dimethyl-1-cyclohexanone
CC1(CCCCC1=O)C
126.2
126.2
2, 2-Dimethylcyclohexanone
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Flammable liquid and vapor.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
Documents

Documents

RUO – Research Use Only