missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,2-Diphenylpropionitrile, 97%, Thermo Scientific Chemicals

$53.33 - $295.84

Chemical Identifiers

CAS 5558-67-8
Molecular Formula C15H13N
Molecular Weight (g/mol) 207.276
MDL Number MFCD00001846
InChI Key DPVHBXFSKLKYIQ-UHFFFAOYSA-N
PubChem CID 79677
IUPAC Name 2,2-diphenylpropanenitrile
SMILES CC(C#N)(C1=CC=CC=C1)C2=CC=CC=C2
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1799106
View Documents
Thermo Scientific Chemicals
A1799106
5 g
Each for $53.33
 
AAA1799109
View Documents
Thermo Scientific Chemicals
A1799109
10 g
Each for $83.26
 
AAA1799118
View Documents
Thermo Scientific Chemicals
A1799118
50 g
Each for $295.84
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

5558-67-8
207.276
DPVHBXFSKLKYIQ-UHFFFAOYSA-N
2,2-diphenylpropanenitrile
C15H13N
MFCD00001846
79677
CC(C#N)(C1=CC=CC=C1)C2=CC=CC=C2

Specifications

5558-67-8
144°C to 145°C (2 mmHg)
C15H13N
MFCD00001846
UN3276
2,2-diphenylpropionitrile, 1,1-diphenylethyl cyanide, 2,2-diphenylpropiononitrile, 2,2-diphenyl propionitrile, .alpha.,.alpha.-diphenylpropionitrile, pubchem12547, acmc-209lnw, 2,2-diphenylpropanitrile, 2,2-diphenyl-propionitrile, 2, 2-diphenyl propionitrile
CC(C#N)(C1=CC=CC=C1)C2=CC=CC=C2
207.276
207.28
2,2-Diphenylpropionitrile
1.108
>110°C (230°F)
1.572
5 g
1955952
DPVHBXFSKLKYIQ-UHFFFAOYSA-N
2,2-diphenylpropanenitrile
79677
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H331-H302-H312
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c

H302+H312+H332

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, LIQUID, TOXIC, N.O.S.

EINECSNumber : 226-925-1

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only