missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,2-Diphenylpropionitrile 98.0+%, TCI America™

$163.92 - $507.57

Chemical Identifiers

CAS 5558-67-8
Molecular Formula C15H13N
Molecular Weight (g/mol) 207.276
MDL Number MFCD00001846
InChI Key DPVHBXFSKLKYIQ-UHFFFAOYSA-N
Synonym 2,2-diphenylpropionitrile, 1,1-diphenylethyl cyanide, 2,2-diphenylpropiononitrile, 2,2-diphenyl propionitrile, .alpha.,.alpha.-diphenylpropionitrile, pubchem12547, acmc-209lnw, 2,2-diphenylpropanitrile, 2,2-diphenyl-propionitrile, 2, 2-diphenyl propionitrile
PubChem CID 79677
IUPAC Name 2,2-diphenylpropanenitrile
SMILES CC(C#N)(C1=CC=CC=C1)C2=CC=CC=C2
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
D25345G
View Documents
TCI America
D25345G
5 g
Each for $163.92
 
D253425G
View Documents
TCI America
D253425G
25 g
Each for $507.57
 
Description

Description

Specifications

Chemical Identifiers

5558-67-8
207.276
DPVHBXFSKLKYIQ-UHFFFAOYSA-N
79677
CC(C#N)(C1=CC=CC=C1)C2=CC=CC=C2
C15H13N
MFCD00001846
2,2-diphenylpropionitrile, 1,1-diphenylethyl cyanide, 2,2-diphenylpropiononitrile, 2,2-diphenyl propionitrile, .alpha.,.alpha.-diphenylpropionitrile, pubchem12547, acmc-209lnw, 2,2-diphenylpropanitrile, 2,2-diphenyl-propionitrile, 2, 2-diphenyl propionitrile
2,2-diphenylpropanenitrile

Specifications

5558-67-8
142°C
MFCD00001846
2,2-diphenylpropionitrile, 1,1-diphenylethyl cyanide, 2,2-diphenylpropiononitrile, 2,2-diphenyl propionitrile, .alpha.,.alpha.-diphenylpropionitrile, pubchem12547, acmc-209lnw, 2,2-diphenylpropanitrile, 2,2-diphenyl-propionitrile, 2, 2-diphenyl propionitrile
CC(C#N)(C1=CC=CC=C1)C2=CC=CC=C2
207.276
207.28
Liquid
Yellow
C15H13N
5 g
DPVHBXFSKLKYIQ-UHFFFAOYSA-N
2,2-diphenylpropanenitrile
79677
≥98.0% (GC)
2,2-Diphenylpropionitrile
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'