missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2,2'-Dithiobis(6-fluorobenzoic Acid) 98.0+%, TCI America™
Supplier: TCI America D31985G
Specifications
2,2′-Dithiobis(6-fluorobenzoic Acid) | |
265°C | |
C14H8F2O4S2 | |
Bis(2-carboxy-3-fluorophenyl) Disulfide | |
C1=CC(=C(C(=C1)SSC2=CC=CC(=C2C(=O)O)F)C(=O)O)F | |
342.331 | |
342.33 | |
Crystalline Powder |
147027-64-3 | |
White-Yellow | |
5 g | |
QEONSZINEMHFPH-UHFFFAOYSA-N | |
2-[(2-carboxy-3-fluorophenyl)disulfanyl]-6-fluorobenzoic acid | |
9974633 | |
≥98.0% (HPLC,T) |
Chemical Identifiers
147027-64-3 | |
342.331 | |
Bis(2-carboxy-3-fluorophenyl) Disulfide | |
2-[(2-carboxy-3-fluorophenyl)disulfanyl]-6-fluorobenzoic acid |
C14H8F2O4S2 | |
QEONSZINEMHFPH-UHFFFAOYSA-N | |
9974633 | |
C1=CC(=C(C(=C1)SSC2=CC=CC(=C2C(=O)O)F)C(=O)O)F |
Safety and Handling
TSCA : No