missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,2'-Dithiodipropionic Acid 98.0+%, TCI America™
$251.65 - $872.38
Chemical Identifiers
| CAS | 4775-93-3 |
|---|---|
| Molecular Formula | C6H10O4S2 |
| Molecular Weight (g/mol) | 210.262 |
| MDL Number | MFCD00059649 |
| InChI Key | UTDRPUGFHHVEDG-UHFFFAOYSA-N |
| PubChem CID | 521224 |
| IUPAC Name | 2-(1-carboxyethyldisulfanyl)propanoic acid |
| SMILES | CC(C(=O)O)SSC(C)C(=O)O |
Chemical Identifiers
| 4775-93-3 | |
| 210.262 | |
| UTDRPUGFHHVEDG-UHFFFAOYSA-N | |
| 2-(1-carboxyethyldisulfanyl)propanoic acid |
| C6H10O4S2 | |
| MFCD00059649 | |
| 521224 | |
| CC(C(=O)O)SSC(C)C(=O)O |
Specifications
| 4775-93-3 | |
| C6H10O4S2 | |
| 5 g | |
| CC(C(=O)O)SSC(C)C(=O)O | |
| 210.262 | |
| 210.26 | |
| Crystalline Powder |
| White-Yellow | |
| MFCD00059649 | |
| UTDRPUGFHHVEDG-UHFFFAOYSA-N | |
| 2-(1-carboxyethyldisulfanyl)propanoic acid | |
| 521224 | |
| ≥98.0% (GC,T) | |
| 2,2′-Dithiodipropionic Acid |
Safety and Handling
TSCA : No