missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-(2-Ethoxyethoxy)ethanol, 99-100%, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor E1022200LTBL

 View more versions of this product

Catalog No. 18608145

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

Specifications

Specifications

0.0001
1
C6H14O3
XXJWXESWEXIICW-UHFFFAOYSA-N
2-(2-ethoxyethoxy)ethan-1-ol
99 to 100%
111-90-0
Drum
200 L
CCOCCOCCO
134.18
Reagent

Chemical Identifiers

111-90-0
134.18
2-(2-ethoxyethoxy)ethan-1-ol
C6H14O3
XXJWXESWEXIICW-UHFFFAOYSA-N
CCOCCOCCO
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.