missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-(2-Hydroxyethyl)-1-methylpyrrolidine 98.0+%, TCI America™
$152.92 - $293.83
Chemical Identifiers
CAS | 67004-64-2 |
---|---|
Molecular Formula | C7H16NO |
Molecular Weight (g/mol) | 130.21 |
MDL Number | MFCD00003174 |
InChI Key | FYVMBPXFPFAECB-ZETCQYMHSA-O |
Synonym | 1-methyl-2-pyrrolidineethanol, 2-1-methylpyrrolidin-2-yl ethanol, 2-pyrrolidineethanol, 1-methyl, 2-2-hydroxyethyl-1-methylpyrrolidine, 2-1-methylpyrrolidin-2-yl ethan-1-ol, 2-1-methyl-2-pyrrolidine ethanol, acmc-209nxf, ksc903s0l, 1-methylpyrrolidine-2-ethanol, 1-methyl-2-pyrrolidine ethanol |
PubChem CID | 93363 |
IUPAC Name | (2S)-2-(2-hydroxyethyl)-1-methylpyrrolidin-1-ium |
SMILES | C[NH+]1CCC[C@H]1CCO |
Chemical Identifiers
67004-64-2 | |
130.21 | |
FYVMBPXFPFAECB-ZETCQYMHSA-O | |
93363 | |
C[NH+]1CCC[C@H]1CCO |
C7H16NO | |
MFCD00003174 | |
1-methyl-2-pyrrolidineethanol, 2-1-methylpyrrolidin-2-yl ethanol, 2-pyrrolidineethanol, 1-methyl, 2-2-hydroxyethyl-1-methylpyrrolidine, 2-1-methylpyrrolidin-2-yl ethan-1-ol, 2-1-methyl-2-pyrrolidine ethanol, acmc-209nxf, ksc903s0l, 1-methylpyrrolidine-2-ethanol, 1-methyl-2-pyrrolidine ethanol | |
(2S)-2-(2-hydroxyethyl)-1-methylpyrrolidin-1-ium |
Specifications
67004-64-2 | |
C7H16NO | |
10 mL | |
FYVMBPXFPFAECB-ZETCQYMHSA-O | |
(2S)-2-(2-hydroxyethyl)-1-methylpyrrolidin-1-ium | |
93363 | |
≥98.0% (GC,T) | |
2-(2-Hydroxyethyl)-1-methylpyrrolidine |
111°C | |
MFCD00003174 | |
1-methyl-2-pyrrolidineethanol, 2-1-methylpyrrolidin-2-yl ethanol, 2-pyrrolidineethanol, 1-methyl, 2-2-hydroxyethyl-1-methylpyrrolidine, 2-1-methylpyrrolidin-2-yl ethan-1-ol, 2-1-methyl-2-pyrrolidine ethanol, acmc-209nxf, ksc903s0l, 1-methylpyrrolidine-2-ethanol, 1-methyl-2-pyrrolidine ethanol | |
C[NH+]1CCC[C@H]1CCO | |
130.21 | |
129.20 | |
Liquid |
Safety and Handling
TSCA : No