missing translation for 'onlineSavingsMsg'
Learn More

2,2'-Isopropylidenebis[(4S)-4-tert-butyl-2-oxazoline], 98%

CAS: 131833-93-7 | C17H30N2O2 | 294.44 g/mol

$284.28 - $955.23

Chemical Identifiers

CAS 131833-93-7
Molecular Formula C17H30N2O2
Molecular Weight (g/mol) 294.44
MDL Number MFCD00192243
InChI Key DPMGLJUMNRDNMX-UHFFFAOYNA-N
Synonym 4s,4's-2,2'-propane-2,2-diyl bis 4-tert-butyl-4,5-dihydrooxazole, s,s---2,2'-isopropylidenebis 4-tert-butyl-2-oxazoline, t-bu-box, s,s, 2,2'-isopropylidenebis 4s-4-tert-butyl-2-oxazoline, s,s---2,2-bis 4-tert-butyl-2-oxazolin-2-yl propane, s,s---2,2-dimethylmethylene bis 4-tert-butyl-2-oxazoline, 4s-4-tert-butyl-2-2-4s-4-tert-butyl-4,5-dihydro-1,3-oxazol-2-yl propan-2-yl-4,5-dihydro-1,3-oxazole, 4-tert-butyl-2-1-4-tert-butyl-4,5-dihydro-1,3-oxazol-2-yl-1-methylethyl-4,5-dihydro-1,3-oxazole, s,s-t-bu-box
PubChem CID 688208
IUPAC Name (4S)-4-tert-butyl-2-[2-[(4S)-4-tert-butyl-4,5-dihydro-1,3-oxazol-2-yl]propan-2-yl]-4,5-dihydro-1,3-oxazole
SMILES CC(C)(C)C1COC(=N1)C(C)(C)C1=NC(CO1)C(C)(C)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC427522500
SDS View Documents
Thermo Scientific Chemicals
427522500
250 mg Glass bottle N/A
Only null left
 
AC427520010
SDS View Documents
Thermo Scientific Chemicals
427520010
1 g Glass bottle N/A
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

131833-93-7
294.44
DPMGLJUMNRDNMX-UHFFFAOYNA-N
688208
CC(C)(C)C1COC(=N1)C(C)(C)C1=NC(CO1)C(C)(C)C
C17H30N2O2
MFCD00192243
4s,4's-2,2'-propane-2,2-diyl bis 4-tert-butyl-4,5-dihydrooxazole, s,s---2,2'-isopropylidenebis 4-tert-butyl-2-oxazoline, t-bu-box, s,s, 2,2'-isopropylidenebis 4s-4-tert-butyl-2-oxazoline, s,s---2,2-bis 4-tert-butyl-2-oxazolin-2-yl propane, s,s---2,2-dimethylmethylene bis 4-tert-butyl-2-oxazoline, 4s-4-tert-butyl-2-2-4s-4-tert-butyl-4,5-dihydro-1,3-oxazol-2-yl propan-2-yl-4,5-dihydro-1,3-oxazole, 4-tert-butyl-2-1-4-tert-butyl-4,5-dihydro-1,3-oxazol-2-yl-1-methylethyl-4,5-dihydro-1,3-oxazole, s,s-t-bu-box
(4S)-4-tert-butyl-2-[2-[(4S)-4-tert-butyl-4,5-dihydro-1,3-oxazol-2-yl]propan-2-yl]-4,5-dihydro-1,3-oxazole

Specifications

131833-93-7
100.0
White to Yellow
Authentic
Glass bottle
MFCD00192243
15, 1297
4s,4's-2,2'-propane-2,2-diyl bis 4-tert-butyl-4,5-dihydrooxazole, s,s---2,2'-isopropylidenebis 4-tert-butyl-2-oxazoline, t-bu-box, s,s, 2,2'-isopropylidenebis 4s-4-tert-butyl-2-oxazoline, s,s---2,2-bis 4-tert-butyl-2-oxazolin-2-yl propane, s,s---2,2-dimethylmethylene bis 4-tert-butyl-2-oxazoline, 4s-4-tert-butyl-2-2-4s-4-tert-butyl-4,5-dihydro-1,3-oxazol-2-yl propan-2-yl-4,5-dihydro-1,3-oxazole, 4-tert-butyl-2-1-4-tert-butyl-4,5-dihydro-1,3-oxazol-2-yl-1-methylethyl-4,5-dihydro-1,3-oxazole, s,s-t-bu-box
CC(C)(C)C1COC(=N1)C(C)(C)C1=NC(CO1)C(C)(C)C
294.44
294.44
Crystalline Powder
98.5
84°C
97.5% min.
98.5% min. (GC)
C17H30N2O2
250 mg
-118° to -122° (20°C, 589nm) (c=5, in CHCL3)
DPMGLJUMNRDNMX-UHFFFAOYNA-N
(4S)-4-tert-butyl-2-[2-[(4S)-4-tert-butyl-4,5-dihydro-1,3-oxazol-2-yl]propan-2-yl]-4,5-dihydro-1,3-oxazole
688208
99%,98% e.e
2, 2'-Isopropylidenebis[(4S)-4-tert-butyl-2-oxazoline], 98% ee
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only