missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-(2-Methyl-1H-imidazol-1-yl)aniline, 97%, Thermo Scientific™

Chemical Identifiers

CAS 26286-55-5
Molecular Formula C10H11N3
Molecular Weight (g/mol) 173.22
MDL Number MFCD08699292
InChI Key LLSZMUSXQITSSJ-UHFFFAOYSA-N
Synonym 2-2-methyl-1h-imidazol-1-yl aniline, 2-2-methyl-imidazol-1-yl-phenylamine, benzenamine,2-2-methyl-1h-imidazol-1-yl, 2-2-methylimidazol-1-yl aniline, 2-2-methylimidazolyl phenylamine, 1-2-amino phenyl-2-methylimidazole, 2-2-methylimidazol-1-yl phenylamine
PubChem CID 13656108
IUPAC Name 2-(2-methylimidazol-1-yl)aniline
SMILES CC1=NC=CN1C1=CC=CC=C1N
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
CC61214CB
SDS View Documents
Thermo Scientific Chemicals
CC61214CB
250mg Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC61214DA
SDS View Documents
Thermo Scientific Chemicals
CC61214DA
1g Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC61214DE
SDS View Documents
Thermo Scientific Chemicals
CC61214DE
5g Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

26286-55-5
173.22
LLSZMUSXQITSSJ-UHFFFAOYSA-N
13656108
CC1=NC=CN1C1=CC=CC=C1N
C10H11N3
MFCD08699292
2-2-methyl-1h-imidazol-1-yl aniline, 2-2-methyl-imidazol-1-yl-phenylamine, benzenamine,2-2-methyl-1h-imidazol-1-yl, 2-2-methylimidazol-1-yl aniline, 2-2-methylimidazolyl phenylamine, 1-2-amino phenyl-2-methylimidazole, 2-2-methylimidazol-1-yl phenylamine
2-(2-methylimidazol-1-yl)aniline

Specifications

26286-55-5
Amber Glass Bottle
MFCD08699292
2-2-methyl-1h-imidazol-1-yl aniline, 2-2-methyl-imidazol-1-yl-phenylamine, benzenamine,2-2-methyl-1h-imidazol-1-yl, 2-2-methylimidazol-1-yl aniline, 2-2-methylimidazolyl phenylamine, 1-2-amino phenyl-2-methylimidazole, 2-2-methylimidazol-1-yl phenylamine
CC1=NC=CN1C1=CC=CC=C1N
173.22
173.22
2-(2-Methyl-1H-imidazol-1-yl)aniline
97%
C10H11N3
250mg
LLSZMUSXQITSSJ-UHFFFAOYSA-N
2-(2-methylimidazol-1-yl)aniline
13656108
97%
Videos
Safety and Handling

Safety and Handling

Warning

SDS
missing translation for 'documents'

missing translation for 'documents'