missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-(2-Methyl-6-nitrophenyl)acetic acid, 95%, Thermo Scientific Chemicals
$91.75 - $263.01
Chemical Identifiers
CAS | 23876-18-8 |
---|---|
Molecular Formula | C9H9NO4 |
Molecular Weight (g/mol) | 195.174 |
MDL Number | MFCD00066313 |
InChI Key | WFKNETIJXXWRHO-UHFFFAOYSA-N |
Synonym | 2-2-methyl-6-nitrophenyl acetic acid, 2-methyl-6-nitrophenylacetic acid, 2-methyl-6-nitrophenyl acetic acid, 2-methyl-6-nitro-phenylacetic acid, 2-6-methyl-2-nitrophenyl acetic acid |
PubChem CID | 675115 |
SMILES | CC1=C(C(=CC=C1)[N+](=O)[O-])CC(=O)O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH66654MD
|
Thermo Scientific Chemicals
H66654MD |
250 mg |
Each for $91.75
|
|
|||||
AAH6665403
|
Thermo Scientific Chemicals
H6665403 |
1 g |
Each for $263.01
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
23876-18-8 | |
195.174 | |
WFKNETIJXXWRHO-UHFFFAOYSA-N | |
675115 |
C9H9NO4 | |
MFCD00066313 | |
2-2-methyl-6-nitrophenyl acetic acid, 2-methyl-6-nitrophenylacetic acid, 2-methyl-6-nitrophenyl acetic acid, 2-methyl-6-nitro-phenylacetic acid, 2-6-methyl-2-nitrophenyl acetic acid | |
CC1=C(C(=CC=C1)[N+](=O)[O-])CC(=O)O |
Specifications
23876-18-8 | |
MFCD00066313 | |
2617187 | |
WFKNETIJXXWRHO-UHFFFAOYSA-N | |
195.174 | |
195.17 | |
Solid |
C9H9NO4 | |
250 mg | |
2-2-methyl-6-nitrophenyl acetic acid, 2-methyl-6-nitrophenylacetic acid, 2-methyl-6-nitrophenyl acetic acid, 2-methyl-6-nitro-phenylacetic acid, 2-6-methyl-2-nitrophenyl acetic acid | |
CC1=C(C(=CC=C1)[N+](=O)[O-])CC(=O)O | |
675115 | |
95% | |
2-(2-Methyl-6-nitrophenyl)acetic acid |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only