Learn More
2,2'-Methylenebis(4-chlorophenol), 95%, Thermo Scientific Chemicals
$67.52 - $359.53
Chemical Identifiers
CAS | 97-23-4 |
---|---|
Molecular Formula | C13H10Cl2O2 |
Molecular Weight (g/mol) | 269.12 |
MDL Number | MFCD00002322 |
InChI Key | MDNWOSOZYLHTCG-UHFFFAOYSA-N |
Synonym | dichlorophen, dichlorophene, dichlorofen, dichlorphen, trivex, 2,2'-methylenebis 4-chlorophenol, didroxan, didroxane, anthiphen, antiphen |
PubChem CID | 3037 |
ChEBI | CHEBI:34689 |
IUPAC Name | 4-chloro-2-[(5-chloro-2-hydroxyphenyl)methyl]phenol |
SMILES | OC1=CC=C(Cl)C=C1CC1=CC(Cl)=CC=C1O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1264014
|
Thermo Scientific Chemicals
A1264014 |
25 g |
N/A
|
|
|||||
AAA1264022
|
Thermo Scientific Chemicals
A1264022 |
100 g |
N/A
|
|
|||||
AAA1264036
|
Thermo Scientific Chemicals
A1264036 |
500 g |
N/A
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
97-23-4 | |
269.12 | |
MDNWOSOZYLHTCG-UHFFFAOYSA-N | |
3037 | |
4-chloro-2-[(5-chloro-2-hydroxyphenyl)methyl]phenol |
C13H10Cl2O2 | |
MFCD00002322 | |
dichlorophen, dichlorophene, dichlorofen, dichlorphen, trivex, 2,2'-methylenebis 4-chlorophenol, didroxan, didroxane, anthiphen, antiphen | |
CHEBI:34689 | |
OC1=CC=C(Cl)C=C1CC1=CC(Cl)=CC=C1O |
Specifications
97-23-4 | |
C13H10Cl2O2 | |
25 g | |
1884514 | |
dichlorophen, dichlorophene, dichlorofen, dichlorphen, trivex, 2,2'-methylenebis 4-chlorophenol, didroxan, didroxane, anthiphen, antiphen | |
OC1=CC=C(Cl)C=C1CC1=CC(Cl)=CC=C1O | |
269.12 | |
CHEBI:34689 | |
95% |
172°C to 176°C | |
MFCD00002322 | |
UN3077 | |
14,3071 | |
MDNWOSOZYLHTCG-UHFFFAOYSA-N | |
4-chloro-2-[(5-chloro-2-hydroxyphenyl)methyl]phenol | |
3037 | |
269.13 | |
2,2'-Methylenebis(4-chlorophenol) |
Safety and Handling
GHS H Statement
H302-H319
Harmful if swallowed.
Causes serious eye irritation.
P264b-P270-P280i-P301+P312-P305+P351+P338-P330-P501c
H302-H319
DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber : 202-567-1
RTECSNumber : SM0175000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only