missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2,2'-O-Cyclocytidine Hydrochloride 98.0+%, TCI America™
Supplier: TCI America C22075G
Specifications
2,2′-O-Cyclocytidine Hydrochloride | |
264°C | |
C9H12ClN3O4 | |
5 g | |
KZOWNALBTMILAP-JBMRGDGGSA-N | |
(2R,4R,5R,6S)-4-(hydroxymethyl)-10-imino-3,7-dioxa-1,9-diazatricyclo[6.4.0.0²,â¶]dodeca-8,11-dien-5-ol hydrochloride | |
74764394 | |
261.66 | |
Crystalline Powder |
10212-25-6 | |
White-Yellow | |
MFCD00012636 | |
cyclocytidine hydrochloride 1g | |
Cl.OC[C@H]1O[C@@H]2[C@@H](OC3=NC(=N)C=CN23)[C@@H]1O | |
261.66 | |
CHEBI:74843 | |
≥98.0% (HPLC,N) |
Chemical Identifiers
10212-25-6 | |
261.66 | |
KZOWNALBTMILAP-JBMRGDGGSA-N | |
74764394 | |
(2R,4R,5R,6S)-4-(hydroxymethyl)-10-imino-3,7-dioxa-1,9-diazatricyclo[6.4.0.0²,â¶]dodeca-8,11-dien-5-ol hydrochloride |
C9H12ClN3O4 | |
MFCD00012636 | |
cyclocytidine hydrochloride 1g | |
CHEBI:74843 | |
Cl.OC[C@H]1O[C@@H]2[C@@H](OC3=NC(=N)C=CN23)[C@@H]1O |
Safety and Handling
RTECSNumber : LV2615000
TSCA : No