missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(2-Thienyl)ethyl p-Toluenesulfonate 95.0+%, TCI America™
$119.30 - $361.62
Chemical Identifiers
| CAS | 40412-06-4 |
|---|---|
| Molecular Formula | C13H14O3S2 |
| Molecular Weight (g/mol) | 282.37 |
| MDL Number | MFCD05150366 |
| InChI Key | HLPRKWVEMYDPAU-UHFFFAOYSA-N |
| Synonym | p-Toluenesulfonic Acid 2-(2-Thienyl)ethyl Ester |
| PubChem CID | 3682034 |
| IUPAC Name | 2-(thiophen-2-yl)ethyl 4-methylbenzene-1-sulfonate |
| SMILES | CC1=CC=C(C=C1)S(=O)(=O)OCCC1=CC=CS1 |
Chemical Identifiers
| 40412-06-4 | |
| 282.37 | |
| HLPRKWVEMYDPAU-UHFFFAOYSA-N | |
| 3682034 | |
| CC1=CC=C(C=C1)S(=O)(=O)OCCC1=CC=CS1 |
| C13H14O3S2 | |
| MFCD05150366 | |
| p-Toluenesulfonic Acid 2-(2-Thienyl)ethyl Ester | |
| 2-(thiophen-2-yl)ethyl 4-methylbenzene-1-sulfonate |
Specifications
| 40412-06-4 | |
| C13H14O3S2 | |
| 5 g | |
| HLPRKWVEMYDPAU-UHFFFAOYSA-N | |
| 2-(thiophen-2-yl)ethyl 4-methylbenzene-1-sulfonate | |
| 3682034 | |
| ≥95.0% (GC) | |
| 2-(2-Thienyl)ethyl p-Toluenesulfonate |
| 32°C | |
| MFCD05150366 | |
| p-Toluenesulfonic Acid 2-(2-Thienyl)ethyl Ester | |
| CC1=CC=C(C=C1)S(=O)(=O)OCCC1=CC=CS1 | |
| 282.37 | |
| 282.37 | |
| Crystalline Powder |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator