missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,3,3-Trimethyl-1-butene, 99+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 140130250

 View more versions of this product

Catalog No. AC140130250

Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2, 3, 3-Trimethyl-1-butene
594-56-9
100.0
0.7000g/mL
-17°C
99+%
C7H14
(CH3)3CC(CH3)=CH2
25 g
0.7
AUYRUAVCWOAHQN-UHFFFAOYSA-N
2,3,3-trimethylbut-1-ene
11669
99+%
99+%
99.0
Colorless
78°C to 80°C
Authentic
Glass bottle
1.4015 to 1.4035
MFCD00008850
01, 221
2,3,3-trimethyl-1-butene, 1-butene, 2,3,3-trimethyl, triptene, acmc-209mdj, 2,3-trimethyl-1-butene, 1-butene,3,3-trimethyl, 1-butene,2,3,3-trimethyl, ch3 3cc ch3 =ch2
CC(=C)C(C)(C)C
98.19
98.19
Liquid

Chemical Identifiers

594-56-9
98.19
AUYRUAVCWOAHQN-UHFFFAOYSA-N
11669
CC(=C)C(C)(C)C
C7H14
MFCD00008850
2,3,3-trimethyl-1-butene, 1-butene, 2,3,3-trimethyl, triptene, acmc-209mdj, 2,3-trimethyl-1-butene, 1-butene,3,3-trimethyl, 1-butene,2,3,3-trimethyl, ch3 3cc ch3 =ch2
2,3,3-trimethylbut-1-ene
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Highly flammable liquid and vapor.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 209-845-1

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only