missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2,3,4,5,6-Pentafluoroanisole, 98%, Thermo Scientific Chemicals
$61.44 - $470.27
Chemical Identifiers
CAS | 389-40-2 |
---|---|
Molecular Formula | C7H3F5O |
Molecular Weight (g/mol) | 198.092 |
MDL Number | MFCD00000289 |
InChI Key | ZRQUIRABLIQJRI-UHFFFAOYSA-N |
Synonym | 2,3,4,5,6-pentafluoroanisole, pentafluoroanisole, benzene, pentafluoromethoxy, pentafluorophenyl methyl ether, pentafluoromethoxybenzene, methyl pentafluorophenyl ether, benzene, 1,2,3,4,5-pentafluoro-6-methoxy, anisole, 2,3,4,5,6-pentafluoro, 1,2,3,4,6-pentafluoro-5-methoxybenzene, methoxypentafluorobenzene |
PubChem CID | 67849 |
IUPAC Name | 1,2,3,4,5-pentafluoro-6-methoxybenzene |
SMILES | COC1=C(C(=C(C(=C1F)F)F)F)F |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAB2392806
|
Thermo Scientific Chemicals
B2392806 |
5 g |
Each for $61.44
|
|
|||||
AAB2392814
|
Thermo Scientific Chemicals
B2392814 |
25 g |
Each for $174.51
|
|
|||||
AAB2392822
|
Thermo Scientific Chemicals
B2392822 |
100 g |
Each for $470.27
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
389-40-2 | |
198.092 | |
ZRQUIRABLIQJRI-UHFFFAOYSA-N | |
67849 | |
COC1=C(C(=C(C(=C1F)F)F)F)F |
C7H3F5O | |
MFCD00000289 | |
2,3,4,5,6-pentafluoroanisole, pentafluoroanisole, benzene, pentafluoromethoxy, pentafluorophenyl methyl ether, pentafluoromethoxybenzene, methyl pentafluorophenyl ether, benzene, 1,2,3,4,5-pentafluoro-6-methoxy, anisole, 2,3,4,5,6-pentafluoro, 1,2,3,4,6-pentafluoro-5-methoxybenzene, methoxypentafluorobenzene | |
1,2,3,4,5-pentafluoro-6-methoxybenzene |
Specifications
389-40-2 | |
1.5 | |
32°C (89°F) | |
1.409 | |
5 g | |
2052664 | |
ZRQUIRABLIQJRI-UHFFFAOYSA-N | |
1,2,3,4,5-pentafluoro-6-methoxybenzene | |
67849 | |
98% |
-38°C | |
136°C to 138°C | |
C7H3F5O | |
MFCD00000289 | |
UN1993 | |
2,3,4,5,6-pentafluoroanisole, pentafluoroanisole, benzene, pentafluoromethoxy, pentafluorophenyl methyl ether, pentafluoromethoxybenzene, methyl pentafluorophenyl ether, benzene, 1,2,3,4,5-pentafluoro-6-methoxy, anisole, 2,3,4,5,6-pentafluoro, 1,2,3,4,6-pentafluoro-5-methoxybenzene, methoxypentafluorobenzene | |
COC1=C(C(=C(C(=C1F)F)F)F)F | |
198.092 | |
198.09 | |
2,3,4,5,6-Pentafluoroanisole |
Safety and Handling
GHS H Statement
H226
Flammable liquid and vapour.
P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
H226
DOTInformation : Hazard Class: 3; Packaging Group: II
EINECSNumber : 206-866-8
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only