missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,3,4,5-Tetrahydro-1H-1,4-benzodiazepine, 95%, Thermo Scientific Chemicals

$70.48 - $199.05

Chemical Identifiers

CAS 5946-39-4
Molecular Formula C9H12N2
Molecular Weight (g/mol) 148.209
MDL Number MFCD03789577
InChI Key MLXBHOCKBUILHN-UHFFFAOYSA-N
Synonym 2,3,4,5-tetrahydro-1h-benzo e 1,4 diazepine, 1h-1,4-benzodiazepine, 2,3,4,5-tetrahydro, 2,3,4,5-tetrahydro-1h benzo e 1,4 diazepine, pubchem14772, acmc-1anok, d0v0nj, tetrahydrobenzo 1,4 diazepine, 2,3,4,5-tetrahydro-1h-1,4-benzodiazepine, 2,3,4,5-tetrahydro-1h-benzo 1,4 diazepine
PubChem CID 2771762
IUPAC Name 2,3,4,5-tetrahydro-1H-1,4-benzodiazepine
SMILES C1CNC2=CC=CC=C2CN1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH27843MD
View Documents
Thermo Scientific Chemicals
H27843MD
250 mg
Each for $70.48
 
AAH2784303
View Documents
Thermo Scientific Chemicals
H2784303
1 g
Each for $199.05
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

5946-39-4
148.209
MLXBHOCKBUILHN-UHFFFAOYSA-N
2771762
C1CNC2=CC=CC=C2CN1
C9H12N2
MFCD03789577
2,3,4,5-tetrahydro-1h-benzo e 1,4 diazepine, 1h-1,4-benzodiazepine, 2,3,4,5-tetrahydro, 2,3,4,5-tetrahydro-1h benzo e 1,4 diazepine, pubchem14772, acmc-1anok, d0v0nj, tetrahydrobenzo 1,4 diazepine, 2,3,4,5-tetrahydro-1h-1,4-benzodiazepine, 2,3,4,5-tetrahydro-1h-benzo 1,4 diazepine
2,3,4,5-tetrahydro-1H-1,4-benzodiazepine

Specifications

5946-39-4
C9H12N2
250 mg
Air Sensitive
MLXBHOCKBUILHN-UHFFFAOYSA-N
2,3,4,5-tetrahydro-1H-1,4-benzodiazepine
2771762
95%
96°C to 100°C
MFCD03789577
UN3259
2,3,4,5-tetrahydro-1h-benzo e 1,4 diazepine, 1h-1,4-benzodiazepine, 2,3,4,5-tetrahydro, 2,3,4,5-tetrahydro-1h benzo e 1,4 diazepine, pubchem14772, acmc-1anok, d0v0nj, tetrahydrobenzo 1,4 diazepine, 2,3,4,5-tetrahydro-1h-1,4-benzodiazepine, 2,3,4,5-tetrahydro-1h-benzo 1,4 diazepine
C1CNC2=CC=CC=C2CN1
148.209
148.21
2,3,4,5-Tetrahydro-1H-1,4-benzodiazepine
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, SOLID, CORROSIVE, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only