missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-(3,4-Dimethylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 98.0+%, TCI America™
$89.19 - $267.56
Chemical Identifiers
CAS | 401797-00-0 |
---|---|
Molecular Formula | C14H21BO2 |
Molecular Weight (g/mol) | 232.13 |
MDL Number | MFCD12405519 |
InChI Key | HWIMTTLCFONAAB-UHFFFAOYSA-N |
Synonym | 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-o-xylene, 3,4-Dimethylphenylboronic Acid Pinacol Ester |
PubChem CID | 15906194 |
IUPAC Name | 2-(3,4-dimethylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane |
SMILES | B1(OC(C(O1)(C)C)(C)C)C2=CC(=C(C=C2)C)C |
Chemical Identifiers
401797-00-0 | |
232.13 | |
HWIMTTLCFONAAB-UHFFFAOYSA-N | |
15906194 | |
B1(OC(C(O1)(C)C)(C)C)C2=CC(=C(C=C2)C)C |
C14H21BO2 | |
MFCD12405519 | |
4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-o-xylene, 3,4-Dimethylphenylboronic Acid Pinacol Ester | |
2-(3,4-dimethylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane |
Specifications
401797-00-0 | |
1 g | |
C14H21BO2 | |
4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-o-xylene, 3,4-Dimethylphenylboronic Acid Pinacol Ester | |
B1(OC(C(O1)(C)C)(C)C)C2=CC(=C(C=C2)C)C | |
232.13 | |
232.13 | |
2-(3,4-Dimethylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane |
White | |
Crystal-Lump at 20°C | |
MFCD12405519 | |
HWIMTTLCFONAAB-UHFFFAOYSA-N | |
2-(3,4-dimethylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane | |
15906194 | |
≥98.0% (GC,T) |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator