missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2,3,5,6-Tetrafluoro-1,4-phenylenediamine 98.0+%, TCI America™
Supplier: TCI America T11101G
Specifications
2,3,5,6-Tetrafluoro-1,4-phenylenediamine | |
145°C | |
MFCD00039767 | |
1,4-Diamino-2,3,5,6-tetrafluorobenzene | |
NC1=C(F)C(F)=C(N)C(F)=C1F | |
180.11 | |
180.11 | |
Crystalline Powder |
1198-64-7 | |
C6H4F4N2 | |
1 g | |
FVFYRXJKYAVFSB-UHFFFAOYSA-N | |
tetrafluorobenzene-1,4-diamine | |
70979 | |
≥98.0% (GC,T) |
Chemical Identifiers
1198-64-7 | |
180.11 | |
FVFYRXJKYAVFSB-UHFFFAOYSA-N | |
70979 | |
NC1=C(F)C(F)=C(N)C(F)=C1F |
C6H4F4N2 | |
MFCD00039767 | |
1,4-Diamino-2,3,5,6-tetrafluorobenzene | |
tetrafluorobenzene-1,4-diamine |
Safety and Handling
TSCA : Yes