missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2,3,5,6-Tetrafluorobenzoic Acid 98.0+%, TCI America™
Supplier: TCI America T18815G
Specifications
2,3,5,6-Tetrafluorobenzoic Acid | |
150°C | |
C7H2F4O2 | |
5 g | |
KVLBXIOFJUWSJQ-UHFFFAOYSA-N | |
2,3,5,6-tetrafluorobenzoic acid | |
567533 | |
≥98.0% (GC,T) |
652-18-6 | |
White-Yellow | |
MFCD00002408 | |
benzoic acid, 2,3,5,6-tetrafluoro, 2,3,5,6-tetrafluorobenzoicacid, pubchem1341, acmc-1avk1, benzoic acid,2,3,5,6-tetrafluoro | |
C1=C(C(=C(C(=C1F)F)C(=O)O)F)F | |
194.085 | |
194.08 | |
Crystalline Powder |
Chemical Identifiers
652-18-6 | |
194.085 | |
KVLBXIOFJUWSJQ-UHFFFAOYSA-N | |
567533 | |
C1=C(C(=C(C(=C1F)F)C(=O)O)F)F |
C7H2F4O2 | |
MFCD00002408 | |
benzoic acid, 2,3,5,6-tetrafluoro, 2,3,5,6-tetrafluorobenzoicacid, pubchem1341, acmc-1avk1, benzoic acid,2,3,5,6-tetrafluoro | |
2,3,5,6-tetrafluorobenzoic acid |
Safety and Handling
TSCA : No