missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2,3,5,6-Tetrafluorobenzyl alcohol, 98%, Thermo Scientific Chemicals
$156.29 - $549.71
Chemical Identifiers
CAS | 4084-38-2 |
---|---|
Molecular Formula | C7H4F4O |
Molecular Weight (g/mol) | 180.10 |
MDL Number | MFCD00792428 |
InChI Key | AGWVQASYTKCTCC-UHFFFAOYSA-N |
PubChem CID | 2734029 |
IUPAC Name | (2,3,5,6-tetrafluorophenyl)methanol |
SMILES | OCC1=C(F)C(F)=CC(F)=C1F |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAB2516206
|
Thermo Scientific Chemicals
B2516206 |
5 g |
Each for $156.29
|
|
|||||
AAB2516214
|
Thermo Scientific Chemicals
B2516214 |
25 g |
Each for $549.71
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
4084-38-2 | |
180.10 | |
AGWVQASYTKCTCC-UHFFFAOYSA-N | |
(2,3,5,6-tetrafluorophenyl)methanol |
C7H4F4O | |
MFCD00792428 | |
2734029 | |
OCC1=C(F)C(F)=CC(F)=C1F |
Specifications
4084-38-2 | |
220°C to 225°C | |
C7H4F4O | |
5 g | |
AGWVQASYTKCTCC-UHFFFAOYSA-N | |
(2,3,5,6-tetrafluorophenyl)methanol | |
2734029 | |
98% |
34°C to 38°C | |
Recognizable | |
MFCD00792428 | |
2,3,5,6-tetrafluorobenzyl alcohol, 2,3,5,6-tetrafluorophenyl methanol, 2,3,5,6-tetrafluorobenzylalcohol, benzenemethanol, 2,3,5,6-tetrafluoro, 2,3,5,6-tetrafluorophenyl methan-1-ol, 2,3,5,6-tetrafluorobenyl alcohol, pubchem15529, acmc-209jh7, 2,3,5,6-tetrafluorbenzyl alcohol, benzenemethanol,2,3,5,6-tetrafluoro | |
OCC1=C(F)C(F)=CC(F)=C1F | |
180.10 | |
180.1 | |
2,3,5,6-Tetrafluorobenzyl alcohol |
Safety and Handling
GHS H Statement
H301-H319-H317
Toxic if swallowed.
Causes serious eye irritation.
May cause an allergic skin reaction.
P261-P264b-P270-P272-P280-P301+P312-P302+P352-P305+P351+P338-P330-P333+P313-P363-P501c
H302-H317-H319
EINECSNumber : 443-840-7
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only