missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-(3,5-Difluorophenyl)acetonitrile, 97%, Thermo Scientific™

Chemical Identifiers

CAS 122376-76-5
Molecular Formula C8H5F2N
Molecular Weight (g/mol) 153.132
MDL Number MFCD00061278
InChI Key OBMYMTSBABEPIB-UHFFFAOYSA-N
Synonym 3,5-difluorophenylacetonitrile, 2-3,5-difluorophenyl acetonitrile, 3,5-difluorobenzyl cyanide, 3,5-difluorobenzylcyanide, benzeneacetonitrile, 3,5-difluoro, 3,5-difluorophenyl acetonitrile, 2-3,5-difluorophenyl ethanenitrile, pubchem1588, acmc-1bxc0
PubChem CID 518565
IUPAC Name 2-(3,5-difluorophenyl)acetonitrile
SMILES C1=C(C=C(C=C1F)F)CC#N
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
SB01685EA
SDS View Documents
Thermo Scientific Chemicals
SB01685EA
10g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

122376-76-5
153.132
OBMYMTSBABEPIB-UHFFFAOYSA-N
518565
C1=C(C=C(C=C1F)F)CC#N
C8H5F2N
MFCD00061278
3,5-difluorophenylacetonitrile, 2-3,5-difluorophenyl acetonitrile, 3,5-difluorobenzyl cyanide, 3,5-difluorobenzylcyanide, benzeneacetonitrile, 3,5-difluoro, 3,5-difluorophenyl acetonitrile, 2-3,5-difluorophenyl ethanenitrile, pubchem1588, acmc-1bxc0
2-(3,5-difluorophenyl)acetonitrile

Specifications

122376-76-5
97%
C8H5F2N
MFCD00061278
3,5-difluorophenylacetonitrile, 2-3,5-difluorophenyl acetonitrile, 3,5-difluorobenzyl cyanide, 3,5-difluorobenzylcyanide, benzeneacetonitrile, 3,5-difluoro, 3,5-difluorophenyl acetonitrile, 2-3,5-difluorophenyl ethanenitrile, pubchem1588, acmc-1bxc0
C1=C(C=C(C=C1F)F)CC#N
153.132
153.13
2-(3, 5-Difluorophenyl)acetonitrile
97.0
Amber Glass Bottle
F2C6H3CH2CN
10g
OBMYMTSBABEPIB-UHFFFAOYSA-N
2-(3,5-difluorophenyl)acetonitrile
518565
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if inhaled.
Causes skin irritation.
May cause respiratory irritation.
Harmful if swallowed.
Causes serious eye irritation.
Harmful in contact with skin.

GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
missing translation for 'documents'

missing translation for 'documents'