missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-(3,5-Dimethyl-1H-pyrazol-1-yl)ethylamine, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H5033303

 View more versions of this product

Catalog No. AAH5033303


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2-(3,5-Dimethyl-1H-pyrazol-1-yl)ethylamine
C7H13N3
1 g
Air and moisture sensitive
FXLOXQBWUPCCDL-UHFFFAOYSA-N
2-(3,5-dimethylpyrazol-1-yl)ethanamine
1091687
97%
62821-88-9
MFCD04967722
UN2735
2-3,5-dimethyl-1h-pyrazol-1-yl ethanamine, 2-3,5-dimethyl-pyrazol-1-yl-ethylamine, 2-3,5-dimethyl-1h-pyrazol-1-yl ethan-1-amine, 2-3,5-dimethylpyrazol-1-yl ethanamine, 2-3,5-dimethylpyrazol-1-yl ethylamine, 2-3,5-dimethyl-1h-pyrazol-1-yl ethylamine, 1-2-aminoethyl-3,5-dimethylpyrazole, 2-3,5-dimethylpyrazolyl ethylamine, acmc-209v2h, 2-3,5-dimethyl-1-pyrazolyl ethanamine
CC1=NN(CCN)C(C)=C1
139.20
139.2

Chemical Identifiers

62821-88-9
139.20
FXLOXQBWUPCCDL-UHFFFAOYSA-N
1091687
C7H13N3
MFCD04967722
2-3,5-dimethyl-1h-pyrazol-1-yl ethanamine, 2-3,5-dimethyl-pyrazol-1-yl-ethylamine, 2-3,5-dimethyl-1h-pyrazol-1-yl ethan-1-amine, 2-3,5-dimethylpyrazol-1-yl ethanamine, 2-3,5-dimethylpyrazol-1-yl ethylamine, 2-3,5-dimethyl-1h-pyrazol-1-yl ethylamine, 1-2-aminoethyl-3,5-dimethylpyrazole, 2-3,5-dimethylpyrazolyl ethylamine, acmc-209v2h, 2-3,5-dimethyl-1-pyrazolyl ethanamine
CC1=NN(CCN)C(C)=C1
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only