missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,3,5-Triphenyl-2H-tetrazolium Chloride Monohydrate,MP Biomedicals
$370.49 - $370.49
Chemical Identifiers
| CAS | 298-96-4 |
|---|---|
| Molecular Formula | C19H15ClN4 |
| Molecular Weight (g/mol) | 334.81 |
| MDL Number | MFCD00011963 |
| InChI Key | PKDBCJSWQUOKDO-UHFFFAOYSA-M |
| Synonym | 2,3,5-triphenyltetrazolium chloride, tetrazolium red, uroscreen, red tetrazolium, urocheck, vitastain, tetrazolium chloride, triphenyltetrazolium chloride, 2,3,5-triphenyl-2h-tetrazolium chloride, tetrazolium chloride |
| PubChem CID | 9283 |
| ChEBI | CHEBI:78019 |
| IUPAC Name | triphenyl-3H-1,2λâµ,3,4-tetrazol-2-ylium chloride |
| SMILES | [Cl-].C1=CC=C(C=C1)N1N=C(N=[N+]1C1=CC=CC=C1)C1=CC=CC=C1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
ICN10312650
|
MP Biomedicals Inc
0210312650 |
50 g |
N/A
|
|
|||||
Chemical Identifiers
| 298-96-4 | |
| 334.81 | |
| PKDBCJSWQUOKDO-UHFFFAOYSA-M | |
| 9283 | |
| triphenyl-3H-1,2λâµ,3,4-tetrazol-2-ylium chloride |
| C19H15ClN4 | |
| MFCD00011963 | |
| 2,3,5-triphenyltetrazolium chloride, tetrazolium red, uroscreen, red tetrazolium, urocheck, vitastain, tetrazolium chloride, triphenyltetrazolium chloride, 2,3,5-triphenyl-2h-tetrazolium chloride, tetrazolium chloride | |
| CHEBI:78019 | |
| [Cl-].C1=CC=C(C=C1)N1N=C(N=[N+]1C1=CC=CC=C1)C1=CC=CC=C1 |
Specifications
| 298-96-4 | |
| C19H15ClN4 | |
| 50 g | |
| PKDBCJSWQUOKDO-UHFFFAOYSA-M | |
| triphenyl-3H-1,2λâµ,3,4-tetrazol-2-ylium chloride | |
| 9283 | |
| 334.81 | |
| 2, 3, 5-Triphenyl-2H-tetrazolium Chloride Monohydrate |
| Tan | |
| MFCD00011963 | |
| 2,3,5-triphenyltetrazolium chloride, tetrazolium red, uroscreen, red tetrazolium, urocheck, vitastain, tetrazolium chloride, triphenyltetrazolium chloride, 2,3,5-triphenyl-2h-tetrazolium chloride, tetrazolium chloride | |
| [Cl-].C1=CC=C(C=C1)N1N=C(N=[N+]1C1=CC=CC=C1)C1=CC=CC=C1 | |
| 334.81 | |
| CHEBI:78019 | |
| Powder |
Safety and Handling
EINECSNumber : 206-071-6
storageNote1 : Protect from light.
Recommended Storage : Store at 4°C.