Learn More
2,3,6,7,10,11-Hexahydroxytriphenylene, 95%, Thermo Scientific Chemicals
$165.19 - $590.32
Chemical Identifiers
CAS | 4877-80-9 |
---|---|
Molecular Formula | C18H12O6 |
Molecular Weight (g/mol) | 324.29 |
InChI Key | QMLILIIMKSKLES-UHFFFAOYSA-N |
Synonym | 2,3,6,7,10,11-hexahydroxytriphenylene, triphenylene-2,3,6,7,10,11-hexaol, 2,3,6,7,10,11-triphenylenehexol, acmc-1ak9f, 2,3,6,7,10,11-hexa-hydroxytriphenylene |
PubChem CID | 11088610 |
IUPAC Name | triphenylene-2,3,6,7,10,11-hexol |
SMILES | C1=C2C3=CC(=C(C=C3C4=CC(=C(C=C4C2=CC(=C1O)O)O)O)O)O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AC457010010
|
Thermo Scientific Chemicals
457010010 |
1 g |
Each for $165.19
|
|
|||||
AC457010050
|
Thermo Scientific Chemicals
457010050 |
5 g |
Each for $590.32
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
4877-80-9 | |
324.29 | |
2,3,6,7,10,11-hexahydroxytriphenylene, triphenylene-2,3,6,7,10,11-hexaol, 2,3,6,7,10,11-triphenylenehexol, acmc-1ak9f, 2,3,6,7,10,11-hexa-hydroxytriphenylene | |
triphenylene-2,3,6,7,10,11-hexol |
C18H12O6 | |
QMLILIIMKSKLES-UHFFFAOYSA-N | |
11088610 | |
C1=C2C3=CC(=C(C=C3C4=CC(=C(C=C4C2=CC(=C1O)O)O)O)O)O |
Specifications
4877-80-9 | |
Authentic | |
Glass Bottle | |
1 g | |
QMLILIIMKSKLES-UHFFFAOYSA-N | |
triphenylene-2,3,6,7,10,11-hexol | |
11088610 | |
95% |
138.0°C | |
94% min. (HPLC) | |
C18H12O6 | |
2,3,6,7,10,11-hexahydroxytriphenylene, triphenylene-2,3,6,7,10,11-hexaol, 2,3,6,7,10,11-triphenylenehexol, acmc-1ak9f, 2,3,6,7,10,11-hexa-hydroxytriphenylene | |
C1=C2C3=CC(=C(C=C3C4=CC(=C(C=C4C2=CC(=C1O)O)O)O)O)O | |
324.29 | |
324.29 | |
2,3,6,7,10,11-Hexahydroxytriphenylene |
Safety and Handling
GHS H Statement Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Call a POISON CENTER or doctor/physician if you feel unwell.
GHS Signal Word: Warning
RUO – Research Use Only