missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,3,6-Trifluorobenzaldehyde, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 384180050

 View more versions of this product

Catalog No. AC384180050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

Specifications

Specifications

2, 3, 6-Trifluorobenzaldehyde
96.0
68°C to 70°C (10.0mmHg)
Authentic
Glass bottle
1.4870 to 1.489
MFCD00061195
benzaldehyde, 2,3,6-trifluoro, 2,3,6-trifluoro-benzaldehyde, pubchem1467, acmc-1bqm6, 2,3,6-trifluorobenzoaldehyde, ksc494g2t, 2,3,6-trifluoro benzaldehyde, timtec-bb sbb003970, attercop-chm at111497
FC1=CC=C(F)C(C=O)=C1F
160.10
160.09
104451-70-9
100.0
81°C
96% min. (GC)
C7H3F3O
F3C6H2CHO
5g
XSBAHBVACIKRTG-UHFFFAOYSA-N
2,3,6-trifluorobenzaldehyde
517845
97%

Chemical Identifiers

104451-70-9
160.10
XSBAHBVACIKRTG-UHFFFAOYSA-N
517845
FC1=CC=C(F)C(C=O)=C1F
C7H3F3O
MFCD00061195
benzaldehyde, 2,3,6-trifluoro, 2,3,6-trifluoro-benzaldehyde, pubchem1467, acmc-1bqm6, 2,3,6-trifluorobenzoaldehyde, ksc494g2t, 2,3,6-trifluoro benzaldehyde, timtec-bb sbb003970, attercop-chm at111497
2,3,6-trifluorobenzaldehyde
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Harmful if swallowed.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only