missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,3-Benzanthracene, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 105260010

 View more versions of this product

Catalog No. AC105260010


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2, 3-Benzanthracene
>300°C
Authentic
Glass bottle
1 g
15,6454
Solubility in water: insoluble. Other solubilities: difficulty soluble in most solvents,soluble in hot xylene
C1=CC=C2C=C3C=C4C=CC=CC4=CC3=CC2=C1
228.29
CHEBI:32600
98%
92-24-0
Orange
97.5% min. (HPLC)
C18H12
05,IV,2545
naphthacene, 2,3-benzanthracene, rubene, benz b anthracene, 2,3-benzanthrene, chrysogen, hydrocarbon, unii-qyj5z6712r, ccris 1183
IFLREYGFSNHWGE-UHFFFAOYSA-N
tetracene
7080
228.29
Crystalline Powder

Chemical Identifiers

92-24-0
228.29
naphthacene, 2,3-benzanthracene, rubene, benz b anthracene, 2,3-benzanthrene, chrysogen, hydrocarbon, unii-qyj5z6712r, ccris 1183
CHEBI:32600
C1=CC=C2C=C3C=C4C=CC=CC4=CC3=CC2=C1
C18H12
IFLREYGFSNHWGE-UHFFFAOYSA-N
7080
tetracene
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Very toxic to aquatic life with long lasting effects.

GHS P Statement
Avoid release to the environment.

GHS Signal Word: Warning

EINECSNumber : 202-138-9

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only