missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2,3-Benzanthracene, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 105265000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
2, 3-Benzanthracene | |
>300°C | |
Authentic | |
Glass bottle | |
500 mg | |
15,6454 | |
Solubility in water: insoluble. Other solubilities: difficulty soluble in most solvents,soluble in hot xylene | |
C1=CC=C2C=C3C=C4C=CC=CC4=CC3=CC2=C1 | |
228.29 | |
CHEBI:32600 | |
98% |
92-24-0 | |
Orange | |
97.5% min. (HPLC) | |
C18H12 | |
05,IV,2545 | |
naphthacene, 2,3-benzanthracene, rubene, benz b anthracene, 2,3-benzanthrene, chrysogen, hydrocarbon, unii-qyj5z6712r, ccris 1183 | |
IFLREYGFSNHWGE-UHFFFAOYSA-N | |
tetracene | |
7080 | |
228.29 | |
Crystalline Powder |
Chemical Identifiers
92-24-0 | |
228.29 | |
naphthacene, 2,3-benzanthracene, rubene, benz b anthracene, 2,3-benzanthrene, chrysogen, hydrocarbon, unii-qyj5z6712r, ccris 1183 | |
CHEBI:32600 | |
C1=CC=C2C=C3C=C4C=CC=CC4=CC3=CC2=C1 |
C18H12 | |
IFLREYGFSNHWGE-UHFFFAOYSA-N | |
7080 | |
tetracene |
Safety and Handling
GHS H Statement
Very toxic to aquatic life with long lasting effects.
GHS P Statement
Avoid release to the environment.
GHS Signal Word: Warning
EINECSNumber : 202-138-9
RUO – Research Use Only