missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-(3-Bromophenyl)-2,3-dihydro-1H-naphtho[1,8-de][1,3,2]diazaborine 98.0+%, TCI America™

$126.40 - $639.90

Chemical Identifiers

CAS 927384-43-8
Molecular Formula C16H12BBrN2
Molecular Weight (g/mol) 323
MDL Number MFCD16038145
InChI Key RDGOAMRKYXEHHS-UHFFFAOYSA-N
Synonym 3-Bromo-1-(2,3-dihydro-1H-naphtho[1,8-de]-1,3,2-diazaborinyl)benzene, 2-(3-Bromophenyl)-2,3-dihydro-1H-naphtho[1,8-de][1,3,2]diazaborinine
PubChem CID 53384412
SMILES B1(NC2=CC=CC3=C2C(=CC=C3)N1)C4=CC(=CC=C4)Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B36541G
View Documents
TCI America
B36541G
1 g
Each for $126.40
 
B36545G
View Documents
TCI America
B36545G
5 g
Each for $639.90
 
Description

Description

Specifications

Chemical Identifiers

927384-43-8
323
RDGOAMRKYXEHHS-UHFFFAOYSA-N
53384412
C16H12BBrN2
MFCD16038145
3-Bromo-1-(2,3-dihydro-1H-naphtho[1,8-de]-1,3,2-diazaborinyl)benzene, 2-(3-Bromophenyl)-2,3-dihydro-1H-naphtho[1,8-de][1,3,2]diazaborinine
B1(NC2=CC=CC3=C2C(=CC=C3)N1)C4=CC(=CC=C4)Br

Specifications

927384-43-8
MFCD16038145
3-Bromo-1-(2,3-dihydro-1H-naphtho[1,8-de]-1,3,2-diazaborinyl)benzene, 2-(3-Bromophenyl)-2,3-dihydro-1H-naphtho[1,8-de][1,3,2]diazaborinine
B1(NC2=CC=CC3=C2C(=CC=C3)N1)C4=CC(=CC=C4)Br
53384412
≥98.0% (GC,N)
2-(3-Bromophenyl)-2,3-dihydro-1H-naphtho[1,8-de][1,3,2]diazaborine
C16H12BBrN2
1 g
RDGOAMRKYXEHHS-UHFFFAOYSA-N
323
323.00
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Refrigerator

SDS
missing translation for 'documents'

missing translation for 'documents'