missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-(3-Bromophenyl)naphthalene, 98%, Thermo Scientific™
$407.45 - $407.45
Chemical Identifiers
CAS | 667940-23-0 |
---|---|
Molecular Formula | C16H11Br |
Molecular Weight (g/mol) | 283.17 |
MDL Number | MFCD16658911 |
InChI Key | FWPXWVYUNHYGPE-UHFFFAOYSA-N |
Synonym | 2-3-bromophenyl naphthalene, naphthalene, 2-3-bromophenyl, 2 3-bromophenyl naphthalene, ksc279g1j |
PubChem CID | 23080773 |
IUPAC Name | 2-(3-bromophenyl)naphthalene |
SMILES | BrC1=CC=CC(=C1)C1=CC2=CC=CC=C2C=C1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH6432206
|
Thermo Scientific Chemicals
H6432206 |
5 g |
Each for $407.45
|
|
|||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
667940-23-0 | |
283.17 | |
FWPXWVYUNHYGPE-UHFFFAOYSA-N | |
23080773 | |
BrC1=CC=CC(=C1)C1=CC2=CC=CC=C2C=C1 |
C16H11Br | |
MFCD16658911 | |
2-3-bromophenyl naphthalene, naphthalene, 2-3-bromophenyl, 2 3-bromophenyl naphthalene, ksc279g1j | |
2-(3-bromophenyl)naphthalene |
Specifications
667940-23-0 | |
MFCD16658911 | |
11441102 | |
FWPXWVYUNHYGPE-UHFFFAOYSA-N | |
2-(3-bromophenyl)naphthalene | |
23080773 | |
98% | |
2-(3-Bromophenyl)naphthalene |
C16H11Br | |
5 g | |
2-3-bromophenyl naphthalene, naphthalene, 2-3-bromophenyl, 2 3-bromophenyl naphthalene, ksc279g1j | |
BrC1=CC=CC(=C1)C1=CC2=CC=CC=C2C=C1 | |
283.17 | |
283.17 | |
Solid |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures