Learn More
2,3-Dibromobutane, (+/-) + meso, 98+%
CAS: 5408-86-6 | C4H8Br2 | 215.916 g/mol
$41.32 - $127.99
Chemical Identifiers
| CAS | 5408-86-6 |
|---|---|
| Molecular Formula | C4H8Br2 |
| Molecular Weight (g/mol) | 215.916 |
| MDL Number | MFCD00000146 |
| InChI Key | BXXWFOGWXLJPPA-UHFFFAOYSA-N |
| Synonym | butane, 2,3-dibromo, dl-2,3-dibromobutane, +/--2,3-dibromobutane, ss-butylene bromide, .+-.-2,3-dibromobutane, dibromo butane, butane,3-dibromo, beta-butylene bromide, r*,r*-+--2,3-dibromobutane, pubchem12534 |
| PubChem CID | 21508 |
| IUPAC Name | 2,3-dibromobutane |
| SMILES | CC(C(C)Br)Br |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1021906
|
Thermo Scientific Chemicals
A1021906 |
5 g |
N/A
|
|
|||||
|
AAA1021914
|
Thermo Scientific Chemicals
A1021914 |
25 g |
N/A
|
|
|||||
|
AAA1021922
|
Thermo Scientific Chemicals
A1021922 |
100 g |
N/A
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 5408-86-6 | |
| 215.916 | |
| BXXWFOGWXLJPPA-UHFFFAOYSA-N | |
| 21508 | |
| CC(C(C)Br)Br |
| C4H8Br2 | |
| MFCD00000146 | |
| butane, 2,3-dibromo, dl-2,3-dibromobutane, +/--2,3-dibromobutane, ss-butylene bromide, .+-.-2,3-dibromobutane, dibromo butane, butane,3-dibromo, beta-butylene bromide, r*,r*-+--2,3-dibromobutane, pubchem12534 | |
| 2,3-dibromobutane |
Specifications
| 5408-86-6 | |
| 166°C to 167°C | |
| C4H8Br2 | |
| MFCD00000146 | |
| 1718916 | |
| BXXWFOGWXLJPPA-UHFFFAOYSA-N | |
| 2,3-dibromobutane | |
| 21508 | |
| ≥98% |
| 1.76 | |
| 123°C (253°F) | |
| 1.5126 | |
| 5 g | |
| butane, 2,3-dibromo, dl-2,3-dibromobutane, +/--2,3-dibromobutane, ss-butylene bromide, .+-.-2,3-dibromobutane, dibromo butane, butane,3-dibromo, beta-butylene bromide, r*,r*-+--2,3-dibromobutane, pubchem12534 | |
| CC(C(C)Br)Br | |
| 215.916 | |
| 215.93 | |
| 2,3-Dibromobutane, (±) + meso |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
H315-H319
DOTInformation : Hazard Class: 6.1; Packaging Group: III
EINECSNumber : 226-476-1
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only