missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,3-Dibromopropene, 80%, tech., stabilized, Thermo Scientific Chemicals

$77.10 - $77.10

Chemical Identifiers

CAS 513-31-5
Molecular Formula C3H4Br2
Molecular Weight (g/mol) 199.87
MDL Number MFCD00000211
InChI Key YMFWYDYJHRGGPF-UHFFFAOYSA-N
Synonym 2,3-dibromopropene, 2,3-dibromo-1-propene, 1-propene, 2,3-dibromo, 2-bromoallyl bromide, 2,3-dibromopropylene, 1,2-dibromo-2-propene, alpha-epidibromohydrin, propene, 2,3-dibromo, unii-8dne7k7z6p, ccris 6712
PubChem CID 10552
IUPAC Name 2,3-dibromoprop-1-ene
SMILES BrCC(Br)=C
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC147740100
SDS View Documents
Thermo Scientific Chemicals
147740100
10 mL Glass bottle
Each for $77.10
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

513-31-5
199.87
YMFWYDYJHRGGPF-UHFFFAOYSA-N
10552
BrCC(Br)=C
C3H4Br2
MFCD00000211
2,3-dibromopropene, 2,3-dibromo-1-propene, 1-propene, 2,3-dibromo, 2-bromoallyl bromide, 2,3-dibromopropylene, 1,2-dibromo-2-propene, alpha-epidibromohydrin, propene, 2,3-dibromo, unii-8dne7k7z6p, ccris 6712
2,3-dibromoprop-1-ene

Specifications

513-31-5
100.0
2.1300g/mL
81°C
75% min. (GC)
C3H4Br2
BrCH2C(Br)=CH2
10 mL
2.13
2,3-dibromopropene, 2,3-dibromo-1-propene, 1-propene, 2,3-dibromo, 2-bromoallyl bromide, 2,3-dibromopropylene, 1,2-dibromo-2-propene, alpha-epidibromohydrin, propene, 2,3-dibromo, unii-8dne7k7z6p, ccris 6712
YMFWYDYJHRGGPF-UHFFFAOYSA-N
2,3-dibromoprop-1-ene
10552
80%
Liquid
75.0
Yellow to Brown
42°C to 44°C (17.0 mmHg)
Authentic
Glass bottle
1.5370 to 1.547
MFCD00000211
01, 201
15, 3030
Solubility in water: insoluble
BrCC(Br)=C
199.87
199.87
Technical
2, 3-Dibromopropene
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.

GHS Signal Word: Danger

EINECSNumber : 208-155-8

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only