missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,3-Dibromopropene 95.0+%, TCI America™

Supplier:  TCI America D020525G

Catalog No. D020525G

Explore available promotions

Description

Description

Specifications

Specifications

2,3-Dibromopropene
Yellow
C3H4Br2
25 g
YMFWYDYJHRGGPF-UHFFFAOYSA-N
2,3-dibromoprop-1-ene
10552
≥95.0% (GC)
513-31-5
141°C
MFCD00000211
2,3-dibromopropene, 2,3-dibromo-1-propene, 1-propene, 2,3-dibromo, 2-bromoallyl bromide, 2,3-dibromopropylene, 1,2-dibromo-2-propene, alpha-epidibromohydrin, propene, 2,3-dibromo, unii-8dne7k7z6p, ccris 6712
BrCC(Br)=C
199.87
199.87
Liquid

Chemical Identifiers

513-31-5
199.87
YMFWYDYJHRGGPF-UHFFFAOYSA-N
10552
BrCC(Br)=C
C3H4Br2
MFCD00000211
2,3-dibromopropene, 2,3-dibromo-1-propene, 1-propene, 2,3-dibromo, 2-bromoallyl bromide, 2,3-dibromopropylene, 1,2-dibromo-2-propene, alpha-epidibromohydrin, propene, 2,3-dibromo, unii-8dne7k7z6p, ccris 6712
2,3-dibromoprop-1-ene
Videos
Safety and Handling

Safety and Handling

RTECSNumber : UC8200000

TSCA : Yes

Recommended Storage : Freezer

SDS
Documents

Documents

Promotions

Promotions