missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dichloro-5-nitropyridine 98.0+%, TCI America™

$54.06 - $214.37

Chemical Identifiers

CAS 22353-40-8
Molecular Formula C5H2Cl2N2O2
Molecular Weight (g/mol) 192.983
MDL Number MFCD03840432
InChI Key XLPDVAVQKGDHNO-UHFFFAOYSA-N
PubChem CID 2763106
IUPAC Name 2,3-dichloro-5-nitropyridine
SMILES C1=C(C=NC(=C1Cl)Cl)[N+](=O)[O-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
D44161G
View Documents
TCI America
D44161G
1 g N/A
Only null left
 
D44165G
View Documents
TCI America
D44165G
5 g N/A
Only null left
 
Description

Description

Specifications

Chemical Identifiers

22353-40-8
192.983
XLPDVAVQKGDHNO-UHFFFAOYSA-N
2,3-dichloro-5-nitropyridine
C5H2Cl2N2O2
MFCD03840432
2763106
C1=C(C=NC(=C1Cl)Cl)[N+](=O)[O-]

Specifications

22353-40-8
Yellow
C5H2Cl2N2O2
1 g
C1=C(C=NC(=C1Cl)Cl)[N+](=O)[O-]
192.983
192.98
Crystalline Powder
53°C
256°C
MFCD03840432
XLPDVAVQKGDHNO-UHFFFAOYSA-N
2,3-dichloro-5-nitropyridine
2763106
≥98.0% (GC)
2,3-Dichloro-5-nitropyridine
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'